Skip to Content
Merck
All Photos(1)

Key Documents

U0508

Sigma-Aldrich

Monoclonal Anti-Ubiquitin antibody produced in mouse

clone 6C1, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6C1, monoclonal

contains

15 mM sodium azide

species reactivity

human, rat, mouse, bovine

technique(s)

indirect ELISA: suitable
microarray: suitable
western blot: 1:1,000 using whole cell lysate of human promyelocitic HL60 cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Ubiquitin, an 8.5kD (approx.) 76 amino acid containing protein, is highly conserved and found in both prokaryotes and eukaryotes. It facilitates protein degradation in ubiquitin/proteasome pathway and also has pivotal role in intracellular regulation. Monoclonal anti-ubiquitin antibody can be used in immunoblotting and ELISA to localize and detect ubiquitin. Mouse anti-ubiquitin antibody reacts specifically with ubiquitin in mouse, human, bovine and rat.

Immunogen

purified bovine ubiquitin conjugated to KLH.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Monoclonal anti-ubiquitin antibody can be used in western blot to visualize and evaluate ubiquitylated proteins.It can also be used in microarray and immunoblotting.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej Susor et al.
Reproduction (Cambridge, England), 134(4), 559-568 (2007-09-25)
In this study, we performed proteomic analysis of porcine oocytes during in vitro maturation. Comparison of oocytes at the initial and final stages of meiotic division characterized candidate proteins that were differentially synthesized during in vitro maturation. While the biosynthesis
Ana P Valencia et al.
BMC musculoskeletal disorders, 18(1), 436-436 (2017-11-11)
Rotator cuff (RTC) tears are a common clinical problem resulting in adverse changes to the muscle, but there is limited information comparing histopathology to contractile function. This study assessed supraspinatus force and susceptibility to injury in the rat model of
Yuga Takahashi et al.
FEBS letters, 585(12), 1741-1747 (2011-05-17)
Nerve growth factor (NGF) binding to its receptor TrkA, which belongs to the family of receptor tyrosine kinases (RTKs), is known to induce its internalization, endosomal trafficking and subsequent lysosomal degradation. The Cbl family of ubiquitin ligases plays a major
Mandy H Y Lam et al.
PloS one, 8(9), e75372-e75372 (2013-09-27)
The yeast HECT-family E3 ubiquitin ligase Rsp5 has been implicated in diverse cell functions. Previously, we and others [1], [2] reported the physical and functional interaction of Rsp5 with the deubiquitinating enzyme Ubp2, and the ubiquitin associated (UBA) domain-containing cofactor
Zi-Yang Zhang et al.
PLoS biology, 17(8), e3000371-e3000371 (2019-08-23)
Inhibitory glycinergic transmission in adult spinal cord is primarily mediated by glycine receptors (GlyRs) containing the α1 subunit. Here, we found that α1ins, a longer α1 variant with 8 amino acids inserted into the intracellular large loop (IL) between transmembrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service