TMO235307
Nunc® low fluorescence sealing tape
polyolefin silicone, non-sterile, case of 100 ea
Synonym(s):
adhesive film, adhesive tape
All Photos(1)
About This Item
UNSPSC Code:
41161502
NACRES:
NB.15
Recommended Products
sterility
non-sterile
packaging
case of 100 ea
manufacturer/tradename
Nunc 235307
Related Categories
General description
“Nunc”Sealing Tapes are effective at reducing evaporation and shielding samples from contamination or accidental spills. These adhesive seals are compatible with various assay applications. Nunc Life Science products guarantee dependable and consistent results, adhering to the highest quality standards and certifications that are reliable within the industry.
Features and Benefits
- Adhesive seals are highly efficient in sealing all microplate formats.
- Pressure sensitive adhesive is not tacky to the touch but creates a solid bond with plates when pressure is applied.
- Thin, lightweight tapes can be applied directly onto the plate using a handheld applicator.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)