Skip to Content
Merck
All Photos(1)

Key Documents

T6704

Sigma-Aldrich

Tandospirone

≥98% (HPLC)

Synonym(s):

(1R*,2S*,3R*,4S*)-N-(4-(4-(2-Pyrimidinyl)-1-piperazinyl)butyl)-2,3-norbornanedicarboximide citrate salt, SM 3997

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29N5O2
CAS Number:
Molecular Weight:
383.49
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

storage condition

protect from light

solubility

DMSO: 38 mg/mL

originator

Dainippon Sumitomo

storage temp.

2-8°C

SMILES string

O=C1[C@@H]2[C@H]3CC[C@H](C3)[C@@H]2C(=O)N1CCCCN4CCN(CC4)c5ncccn5

InChI

1S/C21H29N5O2/c27-19-17-15-4-5-16(14-15)18(17)20(28)26(19)9-2-1-8-24-10-12-25(13-11-24)21-22-6-3-7-23-21/h3,6-7,15-18H,1-2,4-5,8-14H2/t15-,16+,17+,18-

InChI key

CEIJFEGBUDEYSX-FZDBZEDMSA-N

Biochem/physiol Actions

Tandospirone is a partial agonist at 5HT1A receptors. It significantly reduces haloperidol-induced bradykinesia in a dose-dependent manner. The potency of tandospirone is equal to that of buspirone and approximate half that of diazepam. The potency of tandospirone as a dopamine antagonistic is less than 1/4 that of buspirone.

Features and Benefits

This compound was developed by Dainippon Sumitomo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service