Key Documents
T6316
Pharmed tubing
High grade for peristaltic pumps and cell culture, I.D. × O.D. 1/4 in. × 3/8 in.
Synonym(s):
Saint Gobain AY242017, Peristaltic Pump Tubing
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
manufacturer/tradename
Tygon AY242017
I.D. × O.D.
1/4 in. × 3/8 in.
General description
  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Can be autoclaved repeatedly
  • Heat weldable for sterile access
  • Documented biocompatibility to the ISO 109993 standard
  • Meets USP Class VI, FDA, and NSF criteria
Visit the Document Library
Customers Also Viewed
Slide 1 of 1