Skip to Content
Merck
All Photos(1)

Key Documents

T5425

Sigma-Aldrich

Transforming Growth Factor-β3 human

≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, powder, suitable for cell culture

Synonym(s):

hTGF-β3, TGF-β3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Transforming Growth Factor-β3 human, TGF-β3, recombinant, expressed in baculovirus infected Sf21 cells, powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

≥97% (SDS-PAGE)

form

powder

potency

0.01-0.09 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

predicted mol wt ~25 kDa

packaging

pkg of 2 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<0.01 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TGFB3(7043)

Looking for similar products? Visit Product Comparison Guide

General description

The TGFβ3 (transforming Growth Factor-β3) gene is mapped to human chromosome 14q24.3.

Application

Transforming Growth Factor-β3 human has been used to study the role of TGF-β3 in healing tendon-to-bone insertion and in chondrogenic differentiation.

Biochem/physiol Actions

TGF-β (transforming growth factor-β3) isoforms are significantly involved in inducing inflammatory responses. It is associated with vascular diseases, wound healing and ameliorating effect in venous leg ulcer. TGF-β3 has an antagonistic effect on TGF-β1 and reduces the degree of collagen type I deposition and fibroblast differentiation. As a result TGF-β3 might lead to ulcer chronicity due to hindered myofibroblasts formation. TGF-β is a key player in tumor formation and progression. Increased TGF-β3 expression is observed in cancer and adipose tissue fibrosis. TGF-β3 is crucial in tissue regeneration and scarless tissue repair.
TGF-β3 is less prevalent in natural expression than either TGF-β1 or TGF-β2, but it is the most abundant mRNA expressed in chick embryos. It is also expressed in human umbilical cord, in a variety of mesenchymal cells of human and rodent origin, and in several human carcinoma cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in 35% acetonitrile and 0.1% trifluoroacetic acid containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured by its ability to inhibit the IL-4-dependent proliferation of mouse HT-2 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsang, M., et al.
Proc. 7th Intl. Lymphokine Workshop, 9, 607-607 (1990)
The role of transforming growth factor β isoforms in tendon-to-bone healing.
Kim HM, et al.
Connective tissue research, ;52(2), 87-98 (2011)
Roberts, A., and Sporn, M. et al.
Peptide Growth Factors and their Receptors I, 419-472 (1991)
Adipose tissue fibrosis in human cancer cachexia: the role of TGFβ pathway
Alves MJ, et al.
BMC Cancer, 17: 190 (2017)
Chronic Venous Insufficiency: Transforming Growth Factor-β Isoforms and Soluble Endoglin Concentration in Different States of Wound Healing
Ligi D, et al.
International Journal of Molecular Sciences, 18(10) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service