Skip to Content
Merck
All Photos(1)

Key Documents

T5134

Sigma-Aldrich

(±)-α-Tocopherol nicotinate

Synonym(s):

Vitamin E nicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H53NO3
CAS Number:
Molecular Weight:
535.80
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.79

Assay

≥98%

Quality Level

form

solid or liquid

color

white to yellow

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)c3cccnc3)c(C)c(C)c2O1

InChI

1S/C35H53NO3/c1-24(2)13-9-14-25(3)15-10-16-26(4)17-11-20-35(8)21-19-31-29(7)32(27(5)28(6)33(31)39-35)38-34(37)30-18-12-22-36-23-30/h12,18,22-26H,9-11,13-17,19-21H2,1-8H3/t25-,26-,35-/m1/s1

InChI key

MSCCTZZBYHQMQJ-AZAGJHQNSA-N

General description

α-Tocopherol nicotinate is a fat soluble vitamin. It is an ester of tocopherol and niacin. Niacin is linked to the hydroxyl group of active vitamin E in this ester. α-Tocopherol is a form of vitamin E, which is available through dietary sources. RRR-α-tocopherol is a natural form of α-Tocopherol.

Application

(±)-α-Tocopherol nicotinate has been used in metabolic studies to assess its biological functions in rats.

Biochem/physiol Actions

α-Tocopherol is a radical-chain breaker owing to its anti-oxidant property.
Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. α-Tocopherol nicotinate is used along the α-Tocopherol acetate, α-Tocopherol succinate and other esterified forms as a vitamin E supplement with similar but differential activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuru Nakazawa et al.
Ophthalmologica. Journal international d'ophtalmologie. International journal of ophthalmology. Zeitschrift fur Augenheilkunde, 225(2), 120-126 (2010-10-16)
To assess the effects of nilvadipine on the progression of central visual field defect in retinitis pigmentosa (RP). Prospective, randomized, nonmasked, single-center trial. Patients with RP were randomly divided into a treated group receiving oral nilvadipine at 4 mg/day for
N Hidiroglou et al.
Reproduction, nutrition, development, 33(3), 263-268 (1993-01-01)
A 61-d study involving 40 crossbred lambs evaluated the effect of various forms of tocopherol provided daily in equimolar amounts on total cholesterol, high-density lipoprotein-cholesterol and triglyceride concentrations in the serum of lambs. Thirty-five lambs were allotted to 7 treatment
[Effects of soysterol, pantethine and dl-alpha-tocopheryl nicotinate on hyperlipemia in rats (author's transl)].
S Nakayama et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 78(3), 191-202 (1981-09-01)
T W Chung et al.
Diabetic medicine : a journal of the British Diabetic Association, 15(5), 380-385 (1998-06-03)
The effects of alpha-tocopherol nicotinate on blood viscoelasticity and viscosity and on lipid peroxidation stress in erythrocyte membranes in patients with Type 2 DM were investigated. Thirteen Type 2 diabetic subjects with retinopathy were given alpha-tocopherol nicotinate 300 mg tds
K N Prasad et al.
Cancer research, 42(2), 550-555 (1982-02-01)
The effects of various forms of tocopherol (vitamin E) on the growth and differentiation of mouse melanoma (B-16) and mouse fibroblast (L-cells) cells in culture were studied. D-alpha-tocopherol acid succinate induced morphological alterations and growth inhibition in melanoma cells. When

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service