Skip to Content
Merck
All Photos(1)

Key Documents

T3787

Sigma-Aldrich

3,3′,5,5′-Tetraiodothyroacetic acid

≥98% (TLC), powder, thyrointegrin receptor antagonist

Synonym(s):

4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzeneacetic acid, Tetrac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H8O4I4
CAS Number:
Molecular Weight:
747.83
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

product name

3,3′,5,5′-Tetraiodothyroacetic acid,

solubility

acetone: soluble 19.60-20.40 mg/mL, clear, colorless (or faintly yellow to yellow)

Quality Level

storage temp.

−20°C

SMILES string

OC(=O)Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1

InChI

1S/C14H8I4O4/c15-8-4-7(5-9(16)13(8)21)22-14-10(17)1-6(2-11(14)18)3-12(19)20/h1-2,4-5,21H,3H2,(H,19,20)

InChI key

PPJYSSNKSXAVDB-UHFFFAOYSA-N

Application

3,3′,5,5′-Tetraiodothyroacetic acid has been used:
  • as a positive control to study the effects of ioxynil (IOX)
  • diethylstilbestrol (DES) exposure on zebrafish embryos
  • to study its effects on long-term potentiation (LTP) and long-term depression (LTD) in the dentate gyrus in urethane-anesthetized male rats
  • to determine its influence on the actions of thyroid-stimulating hormone
  • thyroid-stimulating immunoglobulins in orbital fibroblast

Biochem/physiol Actions

3,3′,5,5′-Tetraiodothyroacetic acid (Tetrac) is a deaminated analog of L-thyroxine (T4). It prevents the pro-angiogenesis actions of T4 and 3,5,3′-triiodo-L-thyronine. It is a thyrointegrin receptor antagonist. Tetrac prevents the binding of thyroid hormones.
Studies in rats have reported that Tetrac may regulate TSH secretion under in vivo conditions1.

Linkage

Thyroid hormone analog

Preparation Note

3,3′,5,5′-Tetraiodothyroacetic acid is soluble in acetone at 19.60 - 20.40 mg/ml and yields a clear, faint yellow to yellow solution.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul J Davis et al.
Annual review of pharmacology and toxicology, 51, 99-115 (2010-09-28)
Plasma membrane integrin αvβ3 is a cell surface receptor for thyroid hormone at which nongenomic actions are initiated. L-thyroxine (T₄) and 3,3',5-triiodo-L-thyronine (T₃) promote angiogenesis and tumor cell proliferation via the receptor. Tetraiodothyroacetic acid (tetrac), a deaminated T₄ derivative, blocks
Tetrac and NDAT induce Anti-proliferation via Integrin alphavbeta3 in Colorectal Cancers with Different K-RAS Status
Chin YT, et al.
Frontiers in Endocrinology, 10, 130-130 (2019)
Shaker A Mousa et al.
Angiogenesis, 11(2), 183-190 (2007-12-18)
Thyroid hormone has been recently shown to induce tumor growth and angiogenesis via a plasma-membrane hormone receptor on integrin alphaVbeta3. The receptor is at or near the Arg-Gly-Asp (RGD) recognition site on the integrin that is important to extracellular matrix
M Yalcin et al.
The Journal of clinical endocrinology and metabolism, 95(4), 1972-1980 (2010-02-06)
Tetraiodothyroacetic acid (tetrac) blocks angiogenic and tumor cell proliferation actions of thyroid hormone initiated at the cell surface hormone receptor on integrin alphavbeta3. Tetrac also inhibits angiogenesis initiated by vascular endothelial growth factor and basic fibroblast growth factor. We tested
The Effects of Intra-hippocampal L-thyroxine Infusion on Long-term Potentiation and Long-term Depression: A Possible Role for the avb3 Integrin Receptor
Bitiktas S, et al.
Journal of Neuroscience Research (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service