Skip to Content
Merck
All Photos(1)

Key Documents

T3660

Sigma-Aldrich

Calcitonin salmon

≥97% (HPLC), powder

Synonym(s):

Thyrocalcitonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C145H240N44O48S2
CAS Number:
Molecular Weight:
3431.85
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder
Assay:
≥97% (HPLC)

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

solubility

water: 1.00-1.04 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]N[C@H]1CSSC[C@H](NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CO)NC1=O)C(C)O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc2cnc[nH]2)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](C(C)O)C(=O)N5CCC[C@H]5C(N)=O

InChI

1S/C145H240N44O48S2/c1-65(2)45-86(175-139(232)110(70(11)12)183-136(229)99-63-239-238-62-79(148)117(210)178-96(59-191)134(227)174-92(52-104(151)202)131(224)172-90(49-69(9)10)129(222)180-98(61-193)135(228)187-114(74(16)197)142(235)181-99)118(211)158-55-106(204)162-80(25-18-20-40-146)120(213)169-89(48-68(7)8)128(221)179-97(60-192)133(226)167-83(34-37-102(149)200)122(215)165-85(36-39-109(207)208)123(216)171-88(47-67(5)6)127(220)173-91(51-77-54-156-64-161-77)130(223)164-81(26-19-21-41-147)121(214)170-87(46-66(3)4)126(219)166-84(35-38-103(150)201)125(218)186-113(73(15)196)141(234)177-94(50-76-30-32-78(199)33-31-76)143(236)189-44-24-29-101(189)137(230)168-82(27-22-42-157-145(154)155)124(217)185-112(72(14)195)140(233)176-93(53-105(152)203)132(225)184-111(71(13)194)138(231)160-56-107(205)163-95(58-190)119(212)159-57-108(206)182-115(75(17)198)144(237)188-43-23-28-100(188)116(153)209/h30-33,54,64-75,79-101,110-115,190-199H,18-29,34-53,55-63,146-148H2,1-17H3,(H2,149,200)(H2,150,201)(H2,151,202)(H2,152,203)(H2,153,209)(H,156,161)(H,158,211)(H,159,212)(H,160,231)(H,162,204)(H,163,205)(H,164,223)(H,165,215)(H,166,219)(H,167,226)(H,168,230)(H,169,213)(H,170,214)(H,171,216)(H,172,224)(H,173,220)(H,174,227)(H,175,232)(H,176,233)(H,177,234)(H,178,210)(H,179,221)(H,180,222)(H,181,235)(H,182,206)(H,183,229)(H,184,225)(H,185,217)(H,186,218)(H,187,228)(H,207,208)(H4,154,155,157)/t71-,72-,73-,74-,75-,79+,80+,81+,82+,83+,84+,85+,86+,87+,88+,89+,90+,91+,92+,93+,94+,95+,96+,97+,98+,99+,100+,101+,110+,111+,112+,113+,114+,115+/m1/s1

InChI key

BBBFJLBPOGFECG-VJVYQDLKSA-N

Gene Information

human ... CALCR(799)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 [Disulfide bridge: 1-7]

Application

Calcitonin salmon can be used for calcitonin receptor/cAMP assay for assessing the presence of calcitonin receptors expressed by TRAP+ cells.The product can also be used as a test compound for studying as well as comparing the impact of calcitonin gene related peptide (CGRP) and salmon calcitonin (sCT) on gastric mucosal barrier in rats exposed to cold + restraint stress (CRS).

Biochem/physiol Actions

Calcitonin also refers as thyrocalcitonin is a 32 amino acid peptide hormone, which is present abundantly in seminal plasma compare to serum. It acts as a first messenger and hence regulate the the production of cAMP as well as mammalian sperm function. It may also facilitate the regulation of different isoforms of adenylyl cyclase. Additionally, salmon calcitonin stimulates the bone formation and inhibits the bone resorption in postmenopausal osteoporotic women.
Hypocalcemic hormone produced by the parafollicular C cells of the thyroid or by the ultimobranchial bodies of nonmammalian vertebrates. Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.

Other Notes

32 amino acid polypeptide, 8 of which are conserved across all species.
Lyophilized from 0.1% TFA in H2O

Preparation Note

Calcitonin salmon dissolves in 1% acetic acid made in distilled water at a concentration of 1 mg/ml to yield a clear, colorless solution.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K A Kelly et al.
Endocrinology, 139(4), 2092-2101 (1998-04-07)
Stromal cells are required for in vitro osteoclast differentiation and maturation. The murine bone marrow stromally derived BMS2 cell line exhibits adipocytic and osteoblastic features as well as the ability to support lymphopoiesis and myelopoiesis. This work examined the ability
Effects of salmon calcitonin and calcitonin gene related peptide (CGRP) on gastric mucosal barrier in stress induced rats.
Z Kanay
African Journal of Biotechnology, 8 (2009)
M Feigh et al.
British journal of pharmacology, 167(1), 151-163 (2012-04-18)
Oral salmon calcitonin (sCT), a dual-action amylin and calcitonin receptor agonist, improved glucose homeostasis in diet-induced obese rats. Here, we have evaluated the anti-diabetic efficacy of oral sCT using parameters of glycaemic control and beta-cell morphology in male Zucker diabetic
Mathew W Jones et al.
Journal of the American Chemical Society, 134(3), 1847-1852 (2011-12-23)
A series of dibromomaleimides have been shown to be very efficacious at insertion into peptidic disulfide bonds. This conjugation proceeds with a stoichiometric balance of reagents in buffered solutions in less than 15 min to give discrete products while maintaining
Mona M A Abdel-Mottaleb et al.
International journal of pharmaceutics, 515(1-2), 384-389 (2016-11-05)
Recently, a selective preferential accumulation of polymeric nanoparticles (in the size range around 100nm) has been observed in the follicular system of dermatitis skin. The present investigation aimed at clearly investigating the effect of irritant contact dermatitis on the barrier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service