Skip to Content
Merck
All Photos(2)

Key Documents

T3408

Sigma-Aldrich

Tenuazonic acid copper salt from Alternaria alternata

Synonym(s):

3-Acetyl-5-sec-butyl-4-hydroxy-3-pyrrolin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H14NO3 · 1/2Cu
CAS Number:
Molecular Weight:
228.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

CCC(C)C1NC(=O)C(C(C)=O)=C1O[Cu]OC2=C(C(C)=O)C(=O)NC2C(C)CC

InChI

1S/2C10H15NO3.Cu/c2*1-4-5(2)8-9(13)7(6(3)12)10(14)11-8;/h2*5,8,13H,4H2,1-3H3,(H,11,14);/q;;+2/p-2

InChI key

IAHMFKOQYRRWFU-UHFFFAOYSA-L

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steyn, P.S. and Rabie, C.J.,
Phytochemistry, 15, 1977-1977 (1976)
M H Lebrun et al.
Journal of inorganic biochemistry, 24(3), 167-181 (1985-07-01)
Tenuazonic acid (TA) is a phytotoxin produced by a fungal pathogen of rice, Pyricularia oryzae. We have synthesized and characterized the metal complexes of TA with copper (II), iron (III), nickel (II), and magnesium (II). The stoichiometry of the complexes
Stefan Asam et al.
Journal of agricultural and food chemistry, 59(7), 2980-2987 (2011-03-05)
A stable isotope dilution assay (SIDA) for the Alternaria mycotoxin tenuazonic acid was developed. Therefore, [(13)C(6),(15)N]-tenuazonic acid was synthesized from [(13)C(6),(15)N]-isoleucine by Dieckmann intramolecular cyclization after acetoacetylation with diketene. The synthesized [(13)C(6),(15)N]-tenuazonic acid was used as the internal standard for
David Siegel et al.
Analytical and bioanalytical chemistry, 397(2), 453-462 (2009-11-27)
The degradation kinetics of the Alternaria mycotoxin tenuazonic acid (l-TA) in aqueous buffer were studied over a period of 4 months at different pH levels (3.5 and 7.0) and temperatures (4, 25 and 40 degrees C). l-TA and its degradation
David Siegel et al.
Journal of chromatography. A, 1216(21), 4582-4588 (2009-04-14)
Tenuazonic acid (TA) is a major Alternaria mycotoxin. In the present work a novel approach for the detection of TA in cereals by liquid chromatography-ion-trap multistage mass spectrometry after derivatization with 2,4-dinitrophenylhydrazine is described. The product of the derivatization reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service