Skip to Content
Merck
All Photos(1)

Key Documents

T3202

Sigma-Aldrich

Torsemide

≥98% (HPLC), solid

Synonym(s):

Demadex, N-[[(1-Methylethyl)amino]carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N4O3S
CAS Number:
Molecular Weight:
348.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 18 mg/mL
H2O: insoluble

originator

Roche

SMILES string

CC(C)NC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2

InChI

1S/C16H20N4O3S/c1-11(2)18-16(21)20-24(22,23)15-10-17-8-7-14(15)19-13-6-4-5-12(3)9-13/h4-11H,1-3H3,(H,17,19)(H2,18,20,21)

InChI key

NGBFQHCMQULJNZ-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

Application

Torsemide has been used as an inhibitor of Na+ Cl- co-transporter in transgenic Arabidopsis thaliana plants1.

Biochem/physiol Actions

Torsemide functions in the thick ascending limb of the loop of Henle and enhances the excretion of sodium, chloride and water from the luminal side of the cells. Furthermore, torsemide can treat oedematous conditions that are associated with diseases such as liver cirrhosis, kidney disorders and chronic congestive heart failure2.
Torsemide is a loop diuretic of the pyridine-sulfonylurea class with antialdosteronergic properties and inhibitor of the Na+/K+/2Cl- carrier system.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Torsemide is soluble in DMSO at 18 mg/ml and is insoluble in water.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Friedel et al.
Drugs, 41(1), 81-103 (1991-01-01)
Torasemide (torsemide) is a high-ceiling loop diuretic which acts on the thick ascending limb of the loop of Henle to promote rapid and marked excretion of water, sodium and chloride. Like furosemide (frusemide), its major site of action is from
Livia Saleh et al.
Plant signaling & behavior, 8(6), e24259-e24259 (2013-04-23)
The involvement of chloride in salt stress symptoms and salt tolerance mechanisms in plants has been less investigated in the past. Therefore, we studied the salt-induced chloride influx in Arabidopsis expressing the GFP-based anion indicator Clomeleon. High salt concentrations induce
Mark A Oyama et al.
Journal of veterinary cardiology : the official journal of the European Society of Veterinary Cardiology, 13(4), 287-292 (2011-10-28)
Diuretics are a mainstay of therapy in dogs with heart failure. In dogs with advanced heart failure, moderate to high doses of loop diuretics such as furosemide are used with diminishing effects as profound activation of neuroendocrine systems promote signs
J Strobel et al.
Deutsche medizinische Wochenschrift (1946), 136(22), 1207-1210 (2011-05-26)
A 27-year-old woman was admitted to the emergency room because of dyspnoea. Two months earlier she had given birth to a healthy young boy via cesarean section. Computed tomography showed acute embolism in both pulmonary arteries. Chest X-ray and echocardiography
Zarko Jovic et al.
Journal of chromatographic science, 50(4), 324-334 (2012-03-13)
Complete evaluation of chromatographic behavior and establishment of optimal experimental conditions for determination of torasemide and its four impurities are determined by experimental design. Fractional factorial and 3(n) full factorial design were employed for efficient and rapid optimization of liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service