Key Documents
T3168
Nalgene® centrifuge tubes, Oak Ridge Style 3138
nominal capacity 16 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene tubes
About This Item
UNSPSC Code:
41121703
eCl@ss:
32119210
NACRES:
NB.11
Pricing and availability is not currently available.
material
cap (PP sealing cap)
round bottom tube
transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3138-0016
nominal capacity
16 mL
General description
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.
Packaging
With sealing cap
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Jyotirindra Maity et al.
Redox biology, 36, 101622-101622 (2020-08-11)
To define the regulatory role of Kruppel-like factor 2 (KLF2) during osteoblast (OB) differentiation of dental pulp-derived stem cell (DPSC)s, herein, we show that the levels of KLF2 and autophagy-related molecules were significantly increased in differentiated cells. Gain-of-function and loss-of-function