Skip to Content
Merck
All Photos(1)

Key Documents

T2803

Sigma-Aldrich

7-Ethoxy-4-(trifluoromethyl)coumarin

≥98% (TLC), powder

Synonym(s):

Ethyl 4-(trifluoromethyl)umbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9F3O3
CAS Number:
Molecular Weight:
258.19
Beilstein:
8555209
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

7-Ethoxy-4-(trifluoromethyl)coumarin, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

chloroform: 100 mg/mL, clear, colorless (Soluble in chloroform, methanol, and DMSO.)

fluorescence

λex 333 nm; λem 415 nm in methanol

storage temp.

−20°C

SMILES string

CCOc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C12H9F3O3/c1-2-17-7-3-4-8-9(12(13,14)15)6-11(16)18-10(8)5-7/h3-6H,2H2,1H3

InChI key

OLHOIERZAZMHGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

7-Ethoxy-4-(trifluoromethyl)coumarin is a fluorogenic substrate for cytochrome P-450 catalyzed O-de-ethylation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G DeLuca et al.
Biochemical pharmacology, 37(9), 1731-1739 (1988-05-01)
The microsomal O-deethylation of a novel coumarin analog, 7-ethoxy-4-trifluoromethylcoumarin (EFC), to a fluorescent product was characterized. Results indicate that this analog provides a rapid, convenient and highly sensitive means to assay cytochrome P-450-mediated metabolism. Like microsomal 7-ethoxycoumarin (7-EC) O-deethylation, EFC
Thomas Van Leeuwen et al.
Pest management science, 62(5), 425-433 (2006-03-22)
Tetranychus urticae Koch has recently developed resistance to chlorfenapyr in Australia and Japan, but no attempt has yet been made to describe the biochemical mechanisms involved in chlorfenapyr resistance. In this study a laboratory-selected chlorfenapyr-resistant strain was investigated. Resistance to
Maori Mitsuda et al.
Protein expression and purification, 46(2), 401-405 (2005-11-29)
Improvement of CYP2B6 expression was examined by co-expression with molecular chaperones GroES/EL. Although a CO-reduced difference spectrum was not detected in Escherichia coli transformed only by the CYP2B6-expressing vector, co-expression of GroES/EL resulted in high-level expression which reached over 2000
Emily E Scott et al.
Chemical research in toxicology, 15(11), 1407-1413 (2002-11-20)
Until recently, all known structures of bacterial cytochromes P450 suggested that substrate access to the buried active site occurred via the F-G region, a surface loop distal to the heme cavity. However, the structure of P450 51 indicates a large
Shih-Feng Lan et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(4), 1314-1323 (2010-02-23)
In this study, we have evaluated the use of ultra-sterile alginate hydrogels encapsulated with HepG2 liver cells for applications in high throughput drug screening. We have studied the cellular viability and metabolic capacity of the encapsulated cells in two different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service