T2269
L-Tyrosine disodium salt hydrate
≥98% (HPLC), powder
Synonym(s):
L-3-(4-Hydroxyphenyl)alanine disodium salt hydrate
About This Item
Recommended Products
biological source
sugar beets (refined)
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
storage temp.
2-8°C
SMILES string
O.[Na+].[Na+].N[C@@H](Cc1ccc([O-])cc1)C([O-])=O
InChI
1S/C9H11NO3.2Na.H2O/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;;;/h1-4,8,11H,5,10H2,(H,12,13);;;1H2/q;2*+1;/p-2/t8-;;;/m0.../s1
InChI key
SMJHYQCAVHUILN-CZDIJEQGSA-L
Gene Information
human ... GRIN2D(2906)
mouse ... GRIN2D(14814)
rat ... GRIN2D(24412)
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | T1145 | T3754 | T2006 |
---|---|---|---|
powder | powder | powder | powder |
200 | - | 200 | - |
2-8°C | 2-8°C | - | - |
white to beige | white to beige | white to off-white | white to off-white |
sugar beets (refined) | - | - | synthetic (organic) |
Application
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service