Skip to Content
Merck
All Photos(4)

Key Documents

T1580

Sigma-Aldrich

Anti-TDP-43 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ALS10, Anti-TARDBP, Anti-TARDP43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 5-10 μg/mL using rat, mouse, and human kidney
indirect immunofluorescence: 5-10 μg/mL using human HepG2 cells
western blot: 1-2 μg/mL using human U2OS cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARDBP(23435)

Related Categories

General description

TDP-43 (TAR DNA binding protein, TARDP) is a 414 amino acid nuclear protein and is a member of the heterogenous nuclear ribonucleoproteins (hnRNPs) family that bind single stranded RNA. It is encoded by the gene mapped to human chromosome 1p36.22. The encoded protein belongs to the family of heterogenous nuclear ribonucleoproteins (hnRNPs) that bind single stranded RNA. TDP-43 is ubiquitously expressed and is characterized with two RNA-recognition motifs and a glycine-rich C-terminal region.

Specificity

Anti-TDP-43 (C-terminal) specifically recognizes human, mouse, and rat TDP-43.

Biochem/physiol Actions

Heterogeneous nuclear ribonucleoproteins (hnRNPs) play a vital role in generation and processing of RNA, including transcription, splicing, transport and stability. TDP-43 acts as a transcription regulator for human immunodeficiency virus (HIV). Abnormal phosphorylation of TDP-43 at Ser409/410 contributes to the pathology of frontotemporal lobe degeneration subtype (FTLD-U) and amyotrophic lateral sclerosis (ALS).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuriko Katsumata et al.
Acta neuropathologica communications, 6(1), 142-142 (2018-12-21)
TAR-DNA binding protein 43 (TDP-43) proteinopathy is a common brain pathology in elderly persons, but much remains to be learned about this high-morbidity condition. Published stage-based systems for operationalizing disease severity rely on the involvement (presence/absence) of pathology in specific
Clara Bruno et al.
Brain communications, 2(2), fcaa133-fcaa133 (2020-10-03)
Loss-of-function mutations in TANK-binding kinase 1 cause genetic amyotrophic lateral sclerosis and frontotemporal dementia. Consistent with incomplete penetrance in humans, haploinsufficiency of TANK-binding kinase 1 did not cause motor symptoms in mice up to 7 months of age in a previous
Pei-Yi Lin et al.
Journal of neuroscience research, 94(7), 671-682 (2016-03-21)
TAR DNA binding protein 43 (TDP-43) is a nuclear protein that has been shown to have altered homeostasis in the form of neuronal nuclear and cytoplasmic aggregates in some familial and almost all cases of sporadic amyotrophic lateral sclerosis as
The role of TDP-43 in amyotrophic lateral sclerosis and frontotemporal dementia
Mackenzie, Ian RA and Rademakers, Rosa
Current Opinion in Neurology, 21(6), 693-693 (2008)
TDP-43 regulates retinoblastoma protein phosphorylation through the repression of cyclin-dependent kinase 6 expression
Ayala YM, et al.
Proceedings of the National Academy of Sciences, 105(10), 3785-3789 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service