Skip to Content
Merck
All Photos(1)

Key Documents

SRP6257

Sigma-Aldrich

ADIPOQ/Adiponectin/ACRP30 human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Synonym(s):

ACDC, ACRP30, ADIPOQ, APM1, Adiponectin, GBP28

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥92% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 25.4 kDa
observed mol wt 28-33 kDa (DTT-reduced. Protein migrates due to glycosylation. Glu 19 is the predicted N-terminus.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADIPOQ(9370)

General description

Adiponectin (also called Acrp30, AdipoQ) is an adipocyte specific secreted protein that circulates in the plasma. It is induced during adipocyte differentiation and its secretion is stimulated by insulin. The protein has a carboxyl-terminal globular domain and an amino-terminal collagen domain. The gene is mapped to human chromosome 3q27.

Biochem/physiol Actions

Adiponectin (ADIPOQ) has anti-atherosclerotic and anti-inflammatory properties. ADIPOQ has a role in physiological processes such as energy homeostasis and obesity. It exhibits anti-diabetic action by interacting with adiponectin receptors, AdipoR1 and AdipoR2. Low levels of adiponectin are also linked with hypertension and dyslipidemia. Mutation in the gene encoding it is linked with risk for endometrial cancer.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma concentrations of a novel, adipose-specific protein, adiponectin, in type 2 diabetic patients.
Hotta K, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 20(6), 1595-1599 (2000)
Effects of MCF2L2, ADIPOQ and SOX2 genetic polymorphisms on the development of nephropathy in type 1 Diabetes Mellitus.
Zhang D
BMC Medical Genetics, 11, 116-116 (2010)
Adiponectin and adiponectin receptors in insulin resistance, diabetes, and the metabolic syndrome.
Kadowaki T, et al.
The Journal of Clinical Investigation, 116(7), 1784-1792 (2006)
Effects of MCF2L2, ADIPOQ and SOX2 genetic polymorphisms on the development of nephropathy in type 1 Diabetes Mellitus.
Zhang D, et al.
BMC Medical Genetics, 11, 116-116 (2010)
Association Between Single Nucleotide Polymorphism +276G > T (rs1501299) in ADIPOQ and Endometrial Cancer.
Bienkiewicz J, et al.
Pathology Oncology Research, 22(1), 135-138 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service