Skip to Content
Merck
All Photos(1)

Key Documents

SRP5176

Sigma-Aldrich

c-JUN (1-79), GST tagged human

recombinant, expressed in E. coli, ≥50% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AP1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥50% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~35 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... JUN(3725)

General description

c-Jun is a proto-oncogene that forms a complex with c-Fos which regulates transcription from promoters containing AP-1 activation elements. c-Jun has specific DNA binding activity and following in vitro translation, c-Jun binds as a homodimer to the AP-1 DNA site. The transactivating function of c-Jun is acutely regulated by a wide variety of cellular signals via modulation of phosphorylation of two serines (63 and 73). Jun N-terminal domain kinases (JNKs), are responsible for mediating S63/73 phosphorylation on c-Jun in response to a variety of cellular stimuli including TNF-a, heat stress and u.v. light.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T D Halazonetis et al.
Cell, 55(5), 917-924 (1988-12-02)
The c-Jun and c-fos proto-oncogenes encode proteins that form a complex which regulates transcription from promoters containing AP-1 activation elements. c-Jun has specific DNA binding activity, while c-Fos has homology to the putative DNA binding domain of c-Jun. Following in
T Dai et al.
Oncogene, 10(5), 849-855 (1995-03-02)
The transactivating function of the c-Jun proto-oncogene component of the AP-1 transcription factor is acutely regulated by a wide variety of cellular signals via modulation of phosphorylation of two serines (63 and 73). The viral oncoprotein, v-Jun, while containing homologous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service