Skip to Content
Merck
All Photos(1)

Key Documents

SRP3173

Sigma-Aldrich

TIMP-1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

Erythroid-Potentiating activity, Fibroblast collagenase inhibitor, Tissue inhibitor of metalloproteinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

0.5 μg/mL

mol wt

20.6 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

suitability

suitable for molecular biology

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TIMP1(7076)

General description

Research area: Cell SignalingTIMP1 (tissue inhibitor of metalloproteinase 1) is a glycoprotein and belongs to the TIMP family of endogenous MMP (matrix metalloproteinase) inhibitors. Humans contain four TIMPs. All TIMPs contain an N-terminal of 125 residues, a 65-residue C-terminal, and both these domains with three disulfide bonds. The N-terminal domain inhibits MMPs by folding into a separate unit. Recombinant human TIMP-1 is a 20.6 kDa protein containing 184 amino acid residues.

Application

TIMP-1 human has been used for inhibition assays in enzymatic activity assays of Mmp1-CD and Mmp2-CD. It has also been used in interaction immunoassay.

Biochem/physiol Actions

TIMP1 functions as a poor inhibitor of MT1 (membrane type 1)-MMP, MT3-MMP, MT5-MMP and MMP-19. It inhibits ADAM10 (a disintegrin and metalloproteinase domain-containing protein 10) protein. In HUVECs, the down-regulation of TIMP1 and up-regulation of MMP-3 results in aberrant endothelium-dependent vasodilation, EC (endothelial cell) death, and endothelial interruption in a FOXO3 (forkhead box O3)-mediated manner. In patients with CAD (coronary artery disease) and acute coronary syndrome (ACS), the urine levels of this protein are elevated. This protein interacts with Bcl-2 (B cell lymphoma) protein, and induces apoptosis in lung adenocarcinoma cells. The plasma levels of this protein are increased in patients with obesity and obesity-related disorders, such as steatosis, where it participates in pathogenesis of diet-induced hepatic steatosis and glucose intolerance.

Sequence

CTCVPPHPQT AFCNSDLVIR AKFVGTPEVN QTTLYQRYEI KMTKMYKGFQ ALGDAADIRF VYTPAMESVC GYFHRSHNRS EEFLIAGKLQ DGLLHITTCS FVAPWNSLSL AQRRGFTKTY TVGCEECTVF PCLSIPCKLQ SGTHCLWTDQ LLQGSEKGFQ SRHLACLPRE PGLCTWQSLR SQIA

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix metalloproteinase inhibitors as investigative tools in the pathogenesis and management of vascular disease.
Benjamin MM1 and Khalil RA.
EXS, 103, 209-279 (2012)
TIMP-1 Inhibits Apoptosis in Lung Adenocarcinoma Cells via Interaction with Bcl-2.
Nalluri S, et al.
PLoS ONE, 10(9) (2015)
Helena Pulido-Olmo et al.
Frontiers in immunology, 8, 853-853 (2017-08-10)
The protocol describes a novel, rapid, and no-wash one-step immunoassay for highly sensitive and direct detection of the complexes between matrix metalloproteinases (MMPs) and their tissue inhibitor of metalloproteinases (TIMPs) based on AlphaLISA® technology. We describe two procedures: (i) one
Gary J Litherland et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(8), 2175-2187 (2014-04-24)
To assess the role of glycogen synthase kinase 3 (GSK-3) as a regulator of cartilage destruction in human tissue and a murine model of osteoarthritis (OA). Surgical destabilization of the medial meniscus (DMM) was performed to induce experimental murine OA
Tissue Inhibitor Of Matrix Metalloproteinase-1 Is Required for High-Fat Diet-Induced Glucose Intolerance and Hepatic Steatosis in Mice.
Fj?re E, et al.
PLoS ONE, 10(7) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service