Skip to Content
Merck
All Photos(1)

Key Documents

SRP3055

Sigma-Aldrich

Heregulin beta -1 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

NRG1-b1, Neuregulin1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.5 ng/mL

mol wt

7.5 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NRG1(3084)

General description

Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4).[1] HRG1-β1 (heregulin-β1, also referred to as NRG1-β1) contains an Ig (immunoglobulin) domain and an EGF (epidermal growth factor)-like domain that is necessary for direct binding to receptor tyrosine kinases erb3 and erb4.[2][3] The gene HRG1-β1 is mapped to human chromosome 8p12.[4] Recombinant human Heregulin-β1 is a 7.5 kDa polypeptide consisting of only the EGF domain of heregulin-β1 (65 amino acid residues).

Application

Heregulin beta -1 human has been used to enhance proliferation of human dermal fibroblasts (hDFs).[5]
Heregulin-β1 human has been used in the culture medium for the maintenance of Schwann cells.[6] It has been used to study P-Rex1 (PI(3,4,5)P3-dependent Rac exchanger 1)-driven fibroblast invasiveness.[7]

Biochem/physiol Actions

To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 (neuregulin1) gene. Proteolytic processing of the extracellular domain of the transmembrane NRG1 isoforms release soluble growth factors.[8] HRG1-β1 (heregulin-β1, also referred to as NRG1-β1) binding to receptor tyrosine kinases erb3 and erb4 induces erb3 and erb4 heterodimerization with erb2, stimulating intrinsic kinase activity, which leads to tyrosine phosphorylation.[2][9] Although HRG1-β1 biological effects is still unclear, it has been found to promote motility and invasiveness of breast cancer cells which may also involve up-regulation of expression and function of the autocrine motility-promoting factor (AMF).[9]

Sequence

SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The HER2- and heregulin ?1 (HRG)-inducible TNFR superfamily member Fn14 promotes HRG-driven breast cancer cell migration, invasion, and MMP9 expression.
Asrani K
Molecular Cancer Research, 11, 393-404 (2013)
Heregulin-induced activation of ErbB3 by EGFR tyrosine kinase activity promotes tumor growth and metastasis in melanoma cells.
Ueno Y
International Journal of Cancer. Journal International Du Cancer, 123, 340-347 (2008)
Neuregulins: functions, forms, and signaling strategies.
Falls DL
Experimental Cell Research, 284, 14-30 (2003)
Control of peripheral nerve myelination by the beta-secretase BACE1.
Willem M
Science, 314, 664-666 (2006)
Enhancement of wound healing efficiency mediated by artificial dermis functionalized with EGF or NRG1
Yoon D, et al.
Biomedical Materials (Bristol, England), 13(4), 045007-045007 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service