Skip to Content
Merck
All Photos(1)

Key Documents

SRP0291

Sigma-Aldrich

Cathepsin L Active human

recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)

Synonym(s):

CATL, Major excreted protein (MEP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human

recombinant

expressed in FreeStyle 293-F cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

specific activity

≥3900 pmol/min-μg

mol wt

36 kDa

technique(s)

inhibition assay: suitable

suitability

suitable for molecular biology

NCBI accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CTSL(1514)

General description

Research area: Cell Signaling

Cathepsin L is a papain-like cysteine protease and belongs to the Clan A, Family C1. It is composed of L domain of α-helix and an R domain of β-sheet in the spatial structure.

Human cathepsin L (GenBank Accession No. NM_001912), amino acids 18-333, with C-terminal HIS tag, MW = 36 kDa, expressed in FreeStyle 293-F cells.

Application

Active human cathepsin L is useful for the activation of DPP1 (Cathepsin C). Active human cathepsin L has been used:
  • to determine that N-acyl and N-sulfonyloxazolidine-2,4-diones are pseudo-irreversible inhibitors of serine proteases
  • to investigate the role of cathepsin B and L activity in the serum during the human aging process.
  • in inhibitory activity assay
  • to study the role of CTSL in COVID-19 infection

Biochem/physiol Actions

Cathepsin L is a cysteine protease, involved primarily in protein breakdown in the lysosome. Cathepsin L degrades nuclear transcription factors and may affect cell cycle regulation. Cathepsin L is also involved in the immune system, specifically in degrading the invariant chain during major histocompatibility complex (MHC) class II processing, a crucial step in antigen presentation. Its expression in the thymus has been demonstrated to be vital for the development of natural killer cells. Additionally, Cathepsin L contributes to recycling processes during axon outgrowth and synapse formation in the developing postnatal central nervous system. Proteases are associated with the development and progression of cancer. Therefore, cathepsin L may be a potential therapeutic target in cancer treatment.

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1 pmol of substrate per min at 37°C

Physical form

Formulated in 50 mM MES, 400 mM NaCl, pH 5.0, and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Legal Information

FreeStyle is a trademark of Invitrogen Corp.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Ilgová et al.
Molecular and biochemical parasitology, 235, 111248-111248 (2019-12-25)
The gills of the common carp, whose mucosal surface belongs to the key defence mechanisms of piscine immunity, can be infested with both the larval and adult stage of Eudiplozoon nipponicum (Monogenea). Although on their own, monogeneans do not considerably
Ricardo A Orbegozo-Medina et al.
PloS one, 14(2), e0211035-e0211035 (2019-02-02)
Recombinant proteins expressed in E. coli are frequently purified by immobilized metal affinity chromatography (IMAC). By means of this technique, tagged proteins containing a polyhistidine sequence can be obtained up to 95% pure in a single step, but some host
Prohormone Thiol Protease
Vivian Y.H. Hook
Handbook of Proteolytic Enzymes (2013)
Jacqueline M Lankelma et al.
Life sciences, 86(7-8), 225-233 (2009-12-05)
Cathepsin L, a cysteine protease, is considered to be a potential therapeutic target in cancer treatment. Proteases are involved in the development and progression of cancer. Inhibition of activity of specific proteases may slow down cancer progression. In this review
Miao-Miao Zhao et al.
Signal transduction and targeted therapy, 6(1), 134-134 (2021-03-29)
To discover new drugs to combat COVID-19, an understanding of the molecular basis of SARS-CoV-2 infection is urgently needed. Here, for the first time, we report the crucial role of cathepsin L (CTSL) in patients with COVID-19. The circulating level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service