All Photos(1)
About This Item
Linear Formula:
CH3COONH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
form
liquid
Quality Level
application(s)
diagnostic assay manufacturing
shipped in
wet ice
storage temp.
2-8°C
SMILES string
CC([O-])=O.[H][N+]([H])([H])[H]
InChI
1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
InChI key
USFZMSVCRYTOJT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
For Precipitation of DNA and RNA, use 5 M Ammonium Acetate stock soluion to bring the final concentration of ammonium acetate in the working solution to 2.5 M.
Physical form
5M Ammonium Acetate in Water. Solution is 0.2 μm filtered.
Working solution 2.5M to be prepared by dilution with 200 proof Ethanol
Working solution 2.5M to be prepared by dilution with 200 proof Ethanol
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service