Skip to Content
Merck
All Photos(2)

Key Documents

SML3960

Sigma-Aldrich

JNJ-42226314

≥98% (HPLC)

Synonym(s):

[1-(4-Fluorophenyl)indol-5-yl]-[3-[4-(thiazole-2-carbonyl)piperazin-1-yl]azetidin-1-yl]methanone, [4-[1-[[1-(4-Fluorophenyl)-1H-indol-5-yl]carbonyl]-3-azetidinyl]-1-piperazinyl]-2-thiazolylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24FN5O2S
CAS Number:
Molecular Weight:
489.56
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Brain penetrant, highly selective and potent reversible inhibitor of monoacylglycerol lipase (MAGL).


JNJ-42226314 is a brain penetrant, highly selective and potent reversible inhibitor of monoacylglycerol lipase (MAGL)(IC50 = 1.1) that exhibits antinociceptive efficacy in the rat models neuropathic and inflammatory pain. JNJ-42226314 elevates hippocampal 2-arachidonoylglycerol (2-AG) levels and prolongs wakefulness in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The discovery of azetidine-piperazine di-amides as potent, selective and reversible monoacylglycerol lipase (MAGL) inhibitors
Bioorganic & Medicinal Chemistry Letters, 30(14), 127243-127243 (2020)
Ryan M Wyatt et al.
The Journal of pharmacology and experimental therapeutics, 372(3), 339-353 (2019-12-11)
The serine hydrolase monoacylglycerol lipase (MAGL) is the rate-limiting enzyme responsible for the degradation of the endocannabinoid 2-arachidonoylglycerol (2-AG) into arachidonic acid and glycerol. Inhibition of 2-AG degradation leads to elevation of 2-AG, the most abundant endogenous agonist of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service