SML3864
SMG1i
≥95% (HPLC)
Synonym(s):
1-[4-[4-[2-[4-Chloro-3-(diethylsulfamoyl)anilino]pyrimidin-4-yl]pyridin-2-yl]phenyl]-3-methylurea, 2-Chloro-N,N-diethyl-5-[[4-[2-[4-[[(methylamino)carbonyl]amino]phenyl]-4-pyridinyl]-2-pyrimidinyl]amino]benzenesulfonamide, NMDi, SMG1 inhibitor 11j, SMG1 inhibitor compound 11j
About This Item
Quality Level
Assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
SMG1i is a potent and selective SMG-1 kinase inhibitor (hSMG-1 IC50 = 110 pM) with much reduced potency against mTOR, PI3Kα/γ and CDK1/2 (IC50 = 50 nM, 92/60 nM, 32/7.1 μM, respectively). SMG1i selectively downregulates MDA361 cellular phosphorylation of UPF1 (0.3 -1 μM), but not that of mTOR or AKT substrates (p70s6 K and PI3K). Nonsense-mediated decay (NMD) inhibition by SMG1i (1 µM) is shown to synergize with premature termination codons (PTCs) readthrough agents (G418, gentamicin, and paromomycin) in promoting the expression of CFTR protein in murine cells carrying Cftr G542X nonsense mutation.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service