Skip to Content
Merck
All Photos(2)

Key Documents

SML3832

Sigma-Aldrich

Alnustone

≥98% (HPLC)

Synonym(s):

(4E,6E)-1,7-Diphenyl-4,6-heptadien-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O
CAS Number:
Molecular Weight:
262.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCc2ccccc2)\C=C\C=C\c1ccccc1

InChI

1S/C19H18O/c20-19(16-15-18-11-5-2-6-12-18)14-8-7-13-17-9-3-1-4-10-17/h1-14H,15-16H2/b13-7+,14-8+

InChI key

OWMJDOUOHDOUFG-FNCQTZNRSA-N

Biochem/physiol Actions

Alnustone is a diarylheptanoid isolated from Alpinia katsumadai Hayata, rhizomes of Curcuma xanthorrhiza and other plants that exhibits anti-hyperglycemic, anti-inflammatory, antihepatotoxic and antiemetic activities. Alnustone is a potent inhibitor of the Thyroid Adenoma Associated (THADA) function, which reverses THADA-induced β-cell dysfunction and ameliorate hyperglycemia in obese mice. It significantly decreases fed blood glucose and improved glucose tolerance in mice.
Diarylheptanoid isolated from Alpinia katsumadai Hayata and other plants that exhibits anti-hyperglycemic, anti-inflammatory, antihepatotoxic and antiemetic activities

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three non-phenolic diarylheptanoids with anti-inflammatory activity from Curcuma xanthorrhiza
Planta Medica, 59(5), 451-454 (1993)
Alnustone inhibits Streptococcus pneumoniae virulence by targeting pneumolysin and sortase A
Fitoterapia, 162, 105261-105261 (2022)
Yuqing Zhang et al.
Nature communications, 14(1), 1020-1020 (2023-02-24)
Impaired insulin secretion is a hallmark in type 2 diabetes mellitus (T2DM). THADA has been identified as a candidate gene for T2DM, but its role in glucose homeostasis remains elusive. Here we report that THADA is strongly activated in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service