Skip to Content
Merck
All Photos(1)

Key Documents

SML3743

Sigma-Aldrich

Tegobuvir

≥98% (HPLC)

Synonym(s):

GS 9190, 5-({6-[2,4-Bis(trifluoromethyl)phenyl]pyridazin-3-yl}methyl)-2-(2-fluorophenyl)-5H-imidazo[4,5-c]pyridine, 5-[[6-[2,4-Bis(trifluoromethyl)phenyl]-3-pyridazinyl]methyl]-2-(2-fluorophenyl)-5H-imidazo[4,5-c]pyridine, GS 333126, GS-333126, GS-9190, GS333126, GS9190, TGV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H14F7N5
CAS Number:
Molecular Weight:
517.40
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

FC(F)(F)c1c(ccc(c1)C(F)(F)F)c2nnc(cc2)C[n]3cc4nc(nc4cc3)c5c(cccc5)F

InChI

1S/C25H14F7N5/c26-19-4-2-1-3-17(19)23-33-21-9-10-37(13-22(21)34-23)12-15-6-8-20(36-35-15)16-7-5-14(24(27,28)29)11-18(16)25(30,31)32/h1-11,13H,12H2

InChI key

XBEQSQDCBSKCHJ-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, non-cytotoxic, potent and genotype 1 replicons-selective non-nucleoside inhibitor (NNI) against hepatitis C virus (HCV).
Tegobuvir (GS-9190; TGV) is an orally active, non-cytotoxic imidazopyridine that, upon CYP-mediated intracellular activation, forms a glutathione conjugate that acts as a covalent, potent and genotype 1 replicons-selective non-nucleoside inhibitor (NNI) against hepatitis C virus (HCV) NS5B RNA-dependent RNA polymerase (IC50 = 0.8 nM/1b, 13.8 nM/1a, 21.9 μM/2a, >50 μM/1a; IC50 >100 nM against 2b, 3a, 4a, 5a, and 6a). When combined with suboptimal concentrations of other direct acting antivirals (DAAs) in cultures, tegobuvir (6-30 nM) is highly effective in curing cells from HCV replicon and in delaying or preventing the development of resistance against other DAAs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tegobuvir (GS-9190) potency against HCV chimeric replicons derived from consensus NS5B sequences from genotypes 2b, 3a, 4a, 5a, and 6a.
Virology, 429(1), 57-62 (2012)
Mechanistic characterization of GS-9190 (Tegobuvir), a novel nonnucleoside inhibitor of hepatitis C virus NS5B polymerase
Antimicrobial Agents and Chemotherapy, 55(9), 4196-4203 (2011)
Christy M Hebner et al.
PloS one, 7(6), e39163-e39163 (2012-06-22)
Tegobuvir (TGV) is a novel non-nucleoside inhibitor (NNI) of HCV RNA replication with demonstrated antiviral activity in patients with genotype 1 chronic HCV infection. The mechanism of action of TGV has not been clearly defined despite the identification of resistance
Inge Vliegen et al.
Antiviral research, 120, 112-121 (2015-06-04)
Tegobuvir (GS-9190) is a non-nucleoside inhibitor of HCV RNA replication with proven antiviral activity in HCV-infected patients. The in vitro antiviral activity of Tegobuvir, when combined with one or two other direct acting antivirals (DAA) was assessed. When Tegobuvir was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service