Skip to Content
Merck
All Photos(2)

Key Documents

SML3737

Sigma-Aldrich

Octyl-(S)-2HG

≥98% (HPLC)

Synonym(s):

(2S)-2-Hydroxyglutarate octyl ester, (2S)-Octyl-α-hydroxyglutarate, (S)-4-Hydroxy-5-(octyloxy)-5-oxopentanoic acid, 1-Octyl-L-2-hydroxyglutarate, 2S-Hydroxy-pentanedioic acid, 1-octyl ester, L-Octyl-2HG, L2HG, Octyl-(S)-2-hydroxyglutarate, Octyl-L-2HG, S-2HG octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O5
CAS Number:
Molecular Weight:
260.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@@H](CCC(=O)O)C(=O)OCCCCCCCC

InChI

1S/C13H24O5/c1-2-3-4-5-6-7-10-18-13(17)11(14)8-9-12(15)16/h11,14H,2-10H2,1H3,(H,15,16)/t11-/m0/s1

InChI key

UJZOKTKSGUOCCM-NSHDSACASA-N

Biochem/physiol Actions

Membrane-permeant precursor form of the oncometabolite L-2-hydroxyglutarate that inhibits α-ketoglutarate/α-KG-dependent dioxygenases.
Octyl-(S)-2HG (Octyl-L-2HG) is a membrane-permeant precursor form of the oncometabolite L-2-hydroxyglutarate (L2HG) produced from α‐KG by malate dehydrogenases (MDH1/2) and lactate dehydrogenase (LDHA). L2HG can be converted back to α-KG by L-2-hydroxyglutarate dehydrogenase (LHGDH), deletion or mutations of which lead to L2HG accumulation, metabolic disorders and reduced 5hmC levels. Both D- and L-2HG inhibit Jumonji histone demethylases and Tet oxygenases by competing against α-KG binding. However, L2HG is an antagonist, while R2HG (D2HG) is an agonist of α-KG-dependent prolylhydroxylase (EglN).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Wilson et al.
American journal of ophthalmology, 103(2), 167-179 (1987-02-15)
We studied 49 eyes obtained post mortem from 43 patients who had previously had retinal detachment surgery to evaluate some of the factors known to complicate retinal detachment. The retina had been successfully reattached in 44 of 49 eyes. We
Hanna Tarhonskaya et al.
Journal of molecular biology, 429(19), 2895-2906 (2017-08-23)
Methylation of lysine-4 of histone H3 (H3K4men) is an important regulatory factor in eukaryotic transcription. Removal of the transcriptionally activating H3K4 methylation is catalyzed by histone demethylases, including the Jumonji C (JmjC) KDM5 subfamily. The JmjC KDMs are Fe(II) and
Xudong Fu et al.
Cell metabolism, 22(3), 508-515 (2015-07-21)
We discovered recently that the central metabolite α-ketoglutarate (α-KG) extends the lifespan of C. elegans through inhibition of ATP synthase and TOR signaling. Here we find, unexpectedly, that (R)-2-hydroxyglutarate ((R)-2HG), an oncometabolite that interferes with various α-KG-mediated processes, similarly extends
Wanshu Ma et al.
Science advances, 7(18) (2021-05-02)
Recent findings indicate that mitochondrial respiration regulates blood endothelial cell proliferation; however, its role in differentiating lymphatic endothelial cells (LECs) is unknown. We hypothesized that mitochondria could work as a sensor of LECs' metabolic specific needs by determining their functional
Xie Li et al.
Cell metabolism, 35(1), 200-211 (2022-10-30)
Despite its central importance in cellular metabolism, many details remain to be determined regarding subcellular lactate metabolism and its regulation in physiology and disease, as there is sensitive spatiotemporal resolution of lactate distribution, and dynamics remains a technical challenge. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service