Skip to Content
Merck
All Photos(1)

Key Documents

SML3597

Sigma-Aldrich

ARUK3001185

≥98% (HPLC)

Synonym(s):

1-[2,4-Dichloro-3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4Cl2F3N3
CAS Number:
Molecular Weight:
282.05
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

ClC1=C(C(Cl)=CC=C1N2C=CN=N2)C(F)(F)F

InChI

1S/C9H4Cl2F3N3/c10-5-1-2-6(17-4-3-15-16-17)8(11)7(5)9(12,13)14/h1-4H

InChI key

XVFXHCLMGYJAQI-UHFFFAOYSA-N

Biochem/physiol Actions

ARUK3001185 is a brain-penetrant, orally available, potent and selective carboxylesterase Notum inhibitor (IC50 = 6.7 nM) and Wnt signaling activator with no cross-reactivity toward panels of 39 serine hydrolases, 485 kinases (IC50 >10 μM), and 47 human off-target liabilities (IC50 >30 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carboxylesterase Notum Is a Druggable Target to Modulate Wnt Signaling
Journal of Medicinal Chemistry, 64(8), 4289-4311 (2021)
Small-molecule inhibitors of carboxylesterase Notum
Future medicinal chemistry, 13(11), 1001-1015 (2021)
Design of a Potent, Selective, and Brain-Penetrant Inhibitor of Wnt-Deactivating Enzyme Notum by Optimization of a Crystallographic Fragment Hit
Journal of medicinal chemistry, 65(10), 7212-7230 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service