Skip to Content
Merck
All Photos(1)

Key Documents

SML3556

Sigma-Aldrich

Compound 18

≥98% (HPLC)

Synonym(s):

MPO-IN-28, N-(7-Methoxy-4-methyl-2-quinazolinyl)guanidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N5O
CAS Number:
Molecular Weight:
231.25
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N(c1nc2c(c(n1)C)ccc(c2)OC)\C(=N\[H])\N

InChI

1S/C11H13N5O/c1-6-8-4-3-7(17-2)5-9(8)15-11(14-6)16-10(12)13/h3-5H,1-2H3,(H4,12,13,14,15,16)

InChI key

ZJBMSSBTCGJZEE-UHFFFAOYSA-N

Biochem/physiol Actions

Agonist of Aedes aegypti mosquitoes NPY-like receptor 7 (NPYLR7) that inhibit mosquito attraction to humans; potent inhibitor of myeloperoxidase (MPO)
Compound 18 is an agonist of Aedes aegypti mosquitoes NPY-like receptor 7 (NPYLR7) that inhibit mosquito attraction to humans and other hosts. Compound 18 suppresses biting and blood-feeding on a live host. Also, It (MPO-IN-28) is a potent myeloperoxidase (MPO) inhibitor with an IC50 of 44 nM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jalal Soubhye et al.
Journal of medicinal chemistry, 60(15), 6563-6586 (2017-07-04)
The heme enzyme myeloperoxidase (MPO) participates in innate immune defense mechanism through formation of microbicidal reactive oxidants. However, evidence has emerged that MPO-derived oxidants contribute to propagation of inflammatory diseases. Because of the deleterious effects of circulating MPO, there is
Laura B Duvall et al.
Cell, 176(4), 687-701 (2019-02-09)
Female Aedes aegypti mosquitoes bite humans to obtain blood to develop their eggs. Remarkably, their strong attraction to humans is suppressed for days after the blood meal by an unknown mechanism. We investigated a role for neuropeptide Y (NPY)-related signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service