Skip to Content
Merck
All Photos(1)

Key Documents

SML3546

Sigma-Aldrich

KW-2449

≥95% (HPLC)

Synonym(s):

(E)-1-{4-[2-(1H-Indazol-3-yl)vinyl]benzoyl}piperazine, KW 2449, KW2449, [4-[(E)-2-(1H-Indazol-3-yl)ethenyl]phenyl]-1-piperazinylmethanone, [4-[(E)-2-(1H-Indazol-3-yl)ethenyl]phenyl]-piperazin-1-ylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N4O
CAS Number:
Molecular Weight:
332.40
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

KW-2449 is an orally active potent kinase inhibitor against FLT3 (wt/D835Y IC50 = 6.6/1 nM), ABL (wt/T315I IC50 = 14/4 nM), FGFR1 (IC50 = 36 nM), and aurora A (IC50 = 48 nM). KW-2449 also inhibits JAK2, KIT, Src with reduced potency (IC50 = 150, 300, 400 nM, respectively), but not EGFR, Fms, IGF1R or PDGFRα (IC50 >1 μM). In addition to its anti-leukemia efficacy in cultures (GI50 10-650 nM) and in vivo (5-20 mg/kg b.i.d. p.o.), KW-2449 is also a K+/Cl- cotransporter 2 (KCC2) expression-enhancing compound (KEEC) that ameliorates disease phenotypes in a Rett syndrome (RTT) model in Mecp2 mutant mice (2 mg/kg/d i.p.).
Orally active FLT3, ABL, FGFR1, Aurora A inhibitor and a KCC2 expression-enhancing compound (KEEC) with in vivo leukemia & Rett syndrome (RTT) therapeutic efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological enhancement of KCC2 gene expression exerts therapeutic effects on human Rett syndrome neurons and Mecp2 mutant mice
Science Translational Medicine (2019)
KW-2449, a novel multikinase inhibitor, suppresses the growth of leukemia cells with FLT3 mutations or T315I-mutated BCR/ABL translocation
Blood, 114(8), 1607-1617 (2009)
Evaluation of Protein Kinase Inhibitors with PLK4 Cross-Over Potential in a Pre-Clinical Model of Cancer
International Journal of Molecular Sciences, 20 (2019)
HDAC inhibitors potentiate the activity of the BCR/ABL kinase inhibitor KW-2449 in imatinib-sensitive or -resistant BCR/ABL+ leukemia cells in vitro and in viv
Clinical Cancer Research, 17(10), 3219-3232 (2011)
FLT3 ligand impedes the efficacy of FLT3 inhibitors in vitro and in vivo
Blood, 117(12), 3286-3293 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service