Skip to Content
Merck
All Photos(1)

Key Documents

SML3434

Sigma-Aldrich

Erucin

≥98% (HPLC)

Synonym(s):

(4-Isothiocyanatobutyl)(methyl)sulfane; 4-(Methylthio)butyl isothiocyanate, 1-Isothiocyanato-4-(methylthio)butane, ERU, MTBITC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NS2
CAS Number:
Molecular Weight:
161.29
MDL number:
UNSPSC Code:
41121801
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

storage temp.

-10 to -25°C

SMILES string

CSCCCCN=C=S

InChI

1S/C6H11NS2/c1-9-5-3-2-4-7-6-8/h2-5H2,1H3

InChI key

IHQDGXUYTSZGOG-UHFFFAOYSA-N

Biochem/physiol Actions

Erucin (MTBITC) is an orally available, hydrogen sulfide (H2S)-releasing isothiocyanate (ITC) derived from glucoerucin found in the Brassicaceae family of plants (rocket salads), including arugula (Eruca sativa Mill.). Erucin is also formed in vivo through the reduction of sulforaphane, an ITC found in broccoli. Erucin displays neuroprotective, antioxidant, and anticancer efficacy in vitro and in vivo by inducing intracellular reactive oxygen species (ROS) production and Nrf2 activaiton.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piman Pocasap et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 101, 698-709 (2018-03-10)
The structure of the isothiocyanates (ITCs)-erucin, sulforaphane, erysolin, sulforaphene, and phenethyl isothiocyanate-were assessed as well as their respective in vitro anticancer activity on the hepatocellular carcinoma cell line HepG2. All of these ITCs induced both apoptotic and necrotic cell death.
Mohammed Sani Jaafaru et al.
Nutrients, 10(5) (2018-05-09)
Crucifer vegetables, Brassicaceae and other species of the order Brassicales, e.g., Moringaceae that are commonly consumed as spice and food, have been reported to have potential benefits for the treatment and prevention of several health disorders. Though epidemiologically inconclusive, investigations
Umme Salma et al.
Journal of ethnopharmacology, 224, 409-420 (2018-06-19)
Eruca sativa Mill., (Brassicaceae) is a popular remedy for the treatment of hypertension in Pakistan. However, direct effect of the extract and its fractions on blood pressure and vascular tone are unknown. This investigation was aimed to explore the pharmacological
Tadashi Hirata et al.
The Journal of toxicological sciences, 44(2), 73-81 (2019-02-07)
To investigate the chemopreventive mechanisms of 4-Methylthio-3-butenyl isothiocyanate (MTBITC), we analyzed cell viability, cell cycle distribution, and expression levels for cell cycle and apoptosis-related proteins in MTBITC-treated malignant esophageal KYSE510 cells, with and without the reactive oxygen species (ROS) scavenger
Mi-Ok Ko et al.
Journal of microbiology and biotechnology, 26(12), 2036-2042 (2016-09-03)
We evaluated the potentials of 10 isothiocyanates (ITCs) from cruciferous vegetables and radish root hydrolysate for inhibiting the growth of oral pathogens, with an emphasis on assessing any structure-function relationship. Structural differences in ITCs impacted their antimicrobial activities against oral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service