Skip to Content
Merck
All Photos(1)

Key Documents

SML3417

Sigma-Aldrich

Solithromycin

≥95% (HPLC)

Synonym(s):

(3aS,4R,7S,9R,10R,11R,13R,15R,15aR)-1-[4-[4-(3-aminophenyl)-1H-1,2,3-triazol-1-yl]butyl]-4-ethyl-7-fluorooctahydro-11-methoxy-3a,7,9,11,13,15-hexamethyl-10-{[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy}-2H-Oxacyclotetradecino[4,3-d]oxazole-2,6,8,14(1H,7H,9H)-tetrone), CEM 101, OP 1068, OP-1068

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H65FN6O10
CAS Number:
Molecular Weight:
845.01
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C([C@@H](C[C@](C)([C@@H]([C@H](C1=O)C)O[C@]2([C@@H]([C@H](C[C@H](O2)C)N(C)C)O)[H])OC)C)[C@@H]([C@@H]3[C@](C)([C@H](OC([C@@]1(C)F)=O)CC)OC(N3CCCCN4C=C(N=N4)C5=CC(N)=CC=C5)=O)C

InChI

1S/C43H65FN6O10/c1-12-32-43(8)35(50(40(55)60-43)19-14-13-18-49-23-30(46-47-49)28-16-15-17-29(45)21-28)26(4)33(51)24(2)22-41(6,56-11)37(27(5)36(53)42(7,44)39(54)58-32)59-38-34(52)31(48(9)10)20-25(3)57-38/h15-17,21,23-27,31-32,34-35,37-38,52H,12-14,18-20,22,45H2,1-11H3/t24-,25-,26-,27+,31+,32-,34-,35-,37-,38+,41-,42-,43-/m1/s1

InChI key

IXXFZUPTQVDPPK-QIFQIIIXSA-N

Biochem/physiol Actions

Solithromycin is an orally available, broad spectrum, and potent anti-microbial and anti-inflammatory macrolide/fluoroketolide that is effective against respiratory tract pathogens associated with CABP, including macrolide-resistant strains, compared with other available macrolides. Solithromycin significantly inhibits production of TNFα, CXCL8 and MMP9. Also Solithromycin inhibits NF-κB activity under conditions of oxidative stress. Solithromycin exhibits superior anti-microbial and anti-inflammatory activities and is a potential therapy for refractory COPD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiki Kobayashi et al.
The Journal of pharmacology and experimental therapeutics, 345(1), 76-84 (2013-01-30)
Macrolides are reported to reduce exacerbation of chronic inflammatory respiratory disease, such as chronic obstructive pulmonary disease (COPD), and also show anti-inflammatory effects in vitro and in vivo. However the anti-inflammatory efficacies of current macrolides are relatively weak. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service