Skip to Content
Merck
All Photos(1)

Key Documents

SML3386

Sigma-Aldrich

Macitentan

≥98% (HPLC)

Synonym(s):

ACT 064992, ACT-064992, ACT064992, Actelion-1, N-[5-(4-Bromophenyl)-6-[2-[(5-bromo-2-pyrimidinyl)oxy]ethoxy]-4-pyrimidinyl]-N′-propyl-sulfamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20Br2N6O4S
CAS Number:
Molecular Weight:
588.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

BrC1=CC=C(C=C1)C2=C(N=CN=C2OCCOC3=NC=C(C=N3)Br)NS(NCCC)(=O)=O

InChI

1S/C19H20Br2N6O4S/c1-2-7-26-32(28,29)27-17-16(13-3-5-14(20)6-4-13)18(25-12-24-17)30-8-9-31-19-22-10-15(21)11-23-19/h3-6,10-12,26H,2,7-9H2,1H3,(H,24,25,27)

InChI key

JGCMEBMXRHSZKX-UHFFFAOYSA-N

Biochem/physiol Actions

Macitentan (ACT-064992; actelion-1) is an orally active, potent endothelin (ET) receptor antagonist that blocks ET-1-induced cellular calicum response (IC50 in nM = 0.9/HPASMC, 0.8/RASMC, 1.84/m3T3), as well as ET-1-induced contractions of rat aortic rings (ETA) and S6c-induced rat tracheal rings (ETB) ex vivo (pA2 = 7.6 and 5.9, respectively). Macitentan displays therapeutic efficacy in rat hypertension models in vivo (0.3-100 mg/kg p.o. via single or daily dosing).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Treiber et al.
Pharmacology research & perspectives, 8(4), e00619-e00619 (2020-07-03)
The dual endothelin receptor antagonist macitentan was approved in 2013 for the treatment of pulmonary arterial hypertension. Macitentan is an inducer of cytochrome P450 expression in vivo in animal species but not in man. In rat and dog, changes in
Natalia Landázuri et al.
Cells, 10(11) (2021-11-28)
Human cytomegalovirus (HCMV) infection is an important cause of morbidity and mortality in immunocompromised patients and a major etiological factor for congenital birth defects in newborns. Ganciclovir and its pro-drug valganciclovir are the preferred drugs in use today for prophylaxis
Argen Mamazhakypov et al.
Pulmonary circulation, 10(4), 2045894020947283-2045894020947283 (2020-11-27)
Pulmonary arterial hypertension is a severe respiratory disease characterized by pulmonary artery remodeling. RV dysfunction and dysregulated circulating metabolomics are associated with adverse outcomes in pulmonary arterial hypertension. We investigated effects of tadalafil and macitentan alone or in combination on
Soyoung Son et al.
Biomaterials, 276, 121058-121058 (2021-08-17)
Immune checkpoint therapy (ICT), which reinvigorates cytotoxic T cells, provides clinical benefits as an alternative to conventional cancer therapies. However, its clinical response rate is too low to treat an immune-excluded tumor, owing to the presence of abundant stromal elements
Xiaojian Gao et al.
Journal of proteome research, 19(8), 3302-3314 (2020-07-10)
Chronic mountain sickness (CMS) is a high altitude complication with high rates of morbidity and mortality. CMS is characterized by high-altitude polycythemia (HAPC) and high-altitude pulmonary hypertension (HAPH). In this study, macitentan, a dual endothelin receptor antagonist, was used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service