Skip to Content
Merck
All Photos(1)

Key Documents

SML3361

Sigma-Aldrich

Imepitoin

≥98% (HPLC)

Synonym(s):

1-(4-Chlorophenyl)-1,5-dihydro-4-(4-morpholinyl)-2H-imidazol-2-one, 1-(4-Chlorophenyl)-4-morpholin-4-yl-5H-imidazol-2-one, AWD 131-138, AWD131-138, ELB138

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14ClN3O2
CAS Number:
Molecular Weight:
279.72
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N=C(CN1C2=CC=C(C=C2)Cl)N3CCOCC3

InChI

1S/C13H14ClN3O2/c14-10-1-3-11(4-2-10)17-9-12(15-13(17)18)16-5-7-19-8-6-16/h1-4H,5-9H2

InChI key

IQHYCZKIFIHTAI-UHFFFAOYSA-N

Biochem/physiol Actions

Benzodiazepine-binding site GABAA receptor partial positive allosteric modulator (PAM) with in vivo efficacy in animal models of seizure and epilepsy.
Imepitoin (AWD 131-138; ELB 138) is a g-aminobutyric acid A (GABAA) receptor partial positive allosteric modulator (PAM) that targets the benzodiazepine (BZD)-binding site at the α/γ2-interface. Imepitoin enhances GABA current (% response/[GABA] alone in µM/receptor complex = 320/3/α1β2γ2, 310/25/α1β3γ2, 460/8/α2β2γ2, 590/12/α3β2γ2 and 290/3/α5β2γ2 using Xenopus oocytes expressing respective rat receptor complex) with ~2% the potency and 17-27% the efficacy of diazepam toward α1β2γ2. Imepitoin exhibits in vivo therapeutic efficacy in animal models of seizure and epilepsy.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris Rundfeldt et al.
CNS drugs, 28(1), 29-43 (2013-12-21)
Although benzodiazepines (BZDs) offer a wide spectrum of antiepileptic activity against diverse types of epileptic seizures, their use in the treatment of epilepsy is limited because of adverse effects, loss of efficacy (tolerance), and development of physical and psychological dependence.
Wolfgang Löscher et al.
Pharmacological research, 77, 39-46 (2013-09-24)
Recently, the imidazolinone derivative imepitoin has been approved for treatment of canine epilepsy. Imepitoin acts as a low-affinity partial agonist at the benzodiazepine (BZD) site of the GABAA receptor and is the first compound with such mechanism that has been
E Sigel et al.
Neuroscience letters, 245(2), 85-88 (1998-05-30)
Recombinant gamma-aminobutyric acid A (GABA(A)) receptors of the subunit compositions alpha1beta2gamma2, alpha1beta3gamma2, alpha2beta2gamma2, alpha3beta2gamma2 and alpha5beta2gamma2 were expressed in Xenopus oocytes in a functionally active form. At all subunit combinations, AWD 131-138 dose-dependently stimulated GABA currents. At 10 microM AWD
Sevil Yasar et al.
European journal of pharmacology, 465(3), 257-265 (2003-04-19)
AWD 131-138 [1-(4-chlorophenyl)-4-morpholino-imidazolin-2-one], a new low-affinity partial benzodiazepine receptor agonist with potent anticonvulsant and anxiolytic properties in rodent models, was studied in squirrel monkeys trained to discriminate intramuscular (i.m.) injections of midazolam (0.3 mg/kg) from injections of vehicle. Diazepam produced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service