Skip to Content
Merck
All Photos(1)

Key Documents

SML2869

Sigma-Aldrich

Ispinesib

≥98% (HPLC)

Synonym(s):

(R)-N-(3-Aminopropyl)-N-[1-(3-benzyl-7-chloro-4-oxo-3,4-dihydroquinazolin-2-yl)-2-methylpropyl]-4-methylbenzamide, CK0238273, N-(3-Aminopropyl)-N-[(1R)-1-[7-chloro-3,4-dihydro-4-oxo-3-(phenylmethyl)-2-quinazolinyl]-2-methylpropyl]-4-methylbenzamide, SB 715992, SB-715992, SB715992

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H33ClN4O2
CAS Number:
Molecular Weight:
517.06
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +315 to +375°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C30H33ClN4O2/c1-20(2)27(34(17-7-16-32)29(36)23-12-10-21(3)11-13-23)28-33-26-18-24(31)14-15-25(26)30(37)35(28)19-22-8-5-4-6-9-22/h4-6,8-15,18,20,27H,7,16-17,19,32H2,1-3H3/t27-/m1/s1

InChI key

QJZRFPJCWMNVAV-HHHXNRCGSA-N

Biochem/physiol Actions

Ispinesib (SB-715992) is a potent and highly selective kinesin spindle protein (KSP, KIF11, EG5) allosteric inhibtor that specifically inhibits microtubule (MT)-stimulated ATPase activity of KSP (Ki = 1.7 nM; 5 μM MT, 500 μM ATP, 0.75 nM human KSP), but not ubiquitous kinesin heavy chain KHC, neuronal kinesin KIF1A, or mitotic kinesins CENP-E, RabK6, MCAK, MKLP1. Ispinesib exhibits anti-cancer efficacy in cultures (GI50 = 45 nM/BT-474 & 19 nM/MDA-MB-468) and in cancer xenograft models in vivo (8 or 10 mg/kg q4d×3 ip. mice with MCF-7, HCC-1954, KPL4, BT-474 xenografts).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandeep K Talapatra et al.
Journal of medicinal chemistry, 56(16), 6317-6329 (2013-07-24)
Development of drug resistance during cancer chemotherapy is one of the major causes of chemotherapeutic failure for the majority of clinical agents. The aim of this study was to investigate the underlying molecular mechanism of resistance developed by the mitotic
James W Purcell et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(2), 566-576 (2010-01-14)
Ispinesib (SB-715992) is a potent inhibitor of kinesin spindle protein, a kinesin motor protein essential for the formation of a bipolar mitotic spindle and cell cycle progression through mitosis. Clinical studies of ispinesib have shown a 9% response rate in
Lack of efficacy of short term thyroid hormone therapy in Down syndrome.
A P Parikh
Indian pediatrics, 28(1), 83-83 (1991-01-01)
Geng-Yuan Chen et al.
ACS chemical biology, 12(4), 1038-1046 (2017-02-07)
To uncover their contrasting mechanisms, antimitotic drugs that inhibit Eg5 (kinesin-5) were analyzed in mixed-motor gliding assays of kinesin-1 and Eg5 motors in which Eg5 "braking" dominates motility. Loop-5 inhibitors (monastrol, STLC, ispinesib, and filanesib) increased gliding speeds, consistent with
Bojan Milic et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(20), E4613-E4622 (2018-04-29)
Eg5, a mitotic kinesin, has been a target for anticancer drug development. Clinical trials of small-molecule inhibitors of Eg5 have been stymied by the development of resistance, attributable to mitotic rescue by a different endogenous kinesin, KIF15. Compared with Eg5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service