Skip to Content
Merck
All Photos(1)

Key Documents

SML2853

Sigma-Aldrich

DASA-58

≥98% (HPLC)

Synonym(s):

3-(4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-ylsulfonyl)-1,4-diazepan-1-ylsulfonyl)aniline, CID-44543605, DASA 58, DASA58, ML 203, ML-203, ML203, N,N′-DiArylSulfonAmide 58, NCGC00185916

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O6S2
CAS Number:
Molecular Weight:
453.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=CC=CC(S(N2CCN(S(C3=CC=C(OCCO4)C4=C3)(=O)=O)CCC2)(=O)=O)=C1

Biochem/physiol Actions

DASA-58 is a potent and selective activator of cancer-specific pyruvate kinase isoform PKM2 (hPKM2 AC50 = 38 nM, Emax = 82%), but not PKM1, PKR or PKL, that stabilizes PKM2 tetrameric conformation by targeting a site distinct from that of fructose-1,6-bis-phosphate (FBP). DASA-58 exhibits good aqueous solubility (51.2 μg/mL), decreases cellular lactate production and suppresses cell proliferation selectively under hypoxic conditions (30 μM; H1299, 1% O2). When administered in mice, DASA-58 is reported to exhibit anti-thrombotic efficacy in vivo (40 mg/kg via iv).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Monzón-Casanova et al.
Nature immunology, 19(3), 267-278 (2018-01-24)
Antibody affinity maturation occurs in germinal centers (GCs), where B cells cycle between the light zone (LZ) and the dark zone. In the LZ, GC B cells bearing immunoglobulins with the highest affinity for antigen receive positive selection signals from
Qiuyue Kong et al.
Diabetes, 68(2), 361-376 (2018-11-21)
Nonalcoholic steatohepatitis (NASH) is the most prevalent cause of chronic liver disease worldwide. Macrophage-mediated inflammation plays a critical role in NASH pathogenesis; however, optimum therapies for macrophage activation and NASH remain elusive. HSPA12A encodes a novel member of the HSP70
Xinxing Ouyang et al.
Journal of molecular cell biology, 11(2), 93-106 (2018-11-15)
Glucose metabolism plays a key role in thymocyte development. The mammalian target of rapamycin complex 2 (mTORC2) is a critical regulator of cell growth and metabolism, but its role in early thymocyte development and metabolism has not been fully studied.
Matthew B Boxer et al.
Journal of medicinal chemistry, 53(3), 1048-1055 (2009-12-19)
The metabolism of cancer cells is altered to support rapid proliferation. Pharmacological activators of a tumor cell specific pyruvate kinase isozyme (PKM2) may be an approach for altering the classic Warburg effect characteristic of aberrant metabolism in cancer cells yielding
Yang Xu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(11), 12336-12347 (2019-08-28)
Reactive oxygen species (ROS) can act as second messengers in various signaling pathways, and abnormal oxidation contributes to multiple diseases, including cancer. Detecting and quantifying protein oxidation is crucial for a detailed understanding of reduction-oxidation reaction (redox) signaling. We developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service