Skip to Content
Merck
All Photos(1)

Key Documents

SML2482

Sigma-Aldrich

Ticagrelor

≥98% (HPLC)

Synonym(s):

Ticagrelor, (1S,2S,3R,5S)-3-[7-{[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino}-5­(propylthio)-3H-[1,2,3]-triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)cyclopentane-1,2-diol, (1S,2S,3R,5S)-3-[7-{[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino}-5-(propylsulfanyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)-1,2-cyclopentanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28F2N6O4S
CAS Number:
Molecular Weight:
522.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -55 to -65°, c = 0.2 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=C(F)C=C([C@]2([H])C[C@@]2([H])NC3=NC(SCCC)=NC4=C3N=NN4[C@]5([H])C[C@H](OCCO)[C@@H](O)[C@H]5O)C=C1

InChI

1S/C23H28F2N6O4S/c1-2-7-36-23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(35-6-5-32)20(34)19(16)33/h3-4,8,12,15-17,19-20,32-34H,2,5-7,9-10H2,1H3,(H,26,27,28)/t12-,15+,16+,17-,19-,20+/m0/s1

InChI key

OEKWJQXRCDYSHL-FNOIDJSQSA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

General description

Ticagrelor (AZD6140) is a cyclopentyl-triazolopyrimidine.

Biochem/physiol Actions

Ticagrelor is an antithrombotic antiplatelet drug that acts by direct and reversible competitive inhibition of the adensoine purinoceptor P2Y12 receptor. It has clinical benefits in people affected with acute coronary syndrome. It might also play a beneficial role on endothelial function. Ticagrelor can prevent the uptake of adenosine by human erythrocytes and can promote the liberation of adenosine triphosphate (ATP) from human erythrocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michał J Kubisa et al.
Therapeutics and clinical risk management, 14, 129-140 (2018-02-06)
Novel antiplatelet drugs, including ticagrelor, are being successively introduced into the therapy of atherothrombotic conditions due to their superiority over a standard combination of clopidogrel with acetylsalicylic acid in patients with acute coronary syndromes (ACS). A P2Y12 receptor antagonist, ticagrelor
Athanasios Moulias et al.
Journal of cardiovascular pharmacology and therapeutics, 24(1), 11-17 (2018-07-12)
Ticagrelor is a P2Y12 receptor antagonist with proven clinical benefit in patients with acute coronary syndrome. Apart from its principal antiplatelet action, pleiotropic effects have been implicated in the clinical profile of ticagrelor, including a potentially beneficial impact on endothelial
José C Nicolau et al.
Expert opinion on pharmacotherapy, 19(9), 1013-1019 (2018-06-13)
Cardiovascular disease (CVD) is the main cause of death in the world. Coronary artery disease (CAD) is the most common form of CVD presentation, but the prevalence of peripheral artery disease (PAD) is increasing. Patients with polyvascular disease comprise a
Riyaad Aungraheeta et al.
Blood, 128(23), 2717-2728 (2016-10-04)
Ticagrelor is a potent antagonist of the P2Y12 receptor (P2Y12R) and consequently an inhibitor of platelet activity effective in the treatment of atherothrombosis. Here, we sought to further characterize its molecular mechanism of action. Initial studies showed that ticagrelor promoted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service