Skip to Content
Merck
All Photos(1)

Key Documents

SML2346

Sigma-Aldrich

Ganciclovir

≥99% (HPLC), DMSO solution, viral DNA elongation inhibitor

Synonym(s):

Ganciclovir ready made solution, 2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, 9-(1,3-Dihydroxy-2-propoxymethyl)guanine, DHPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5O4
CAS Number:
Molecular Weight:
255.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Ganciclovir ready made solution, ≥99% (HPLC), 10 mM in DMSO

Assay

≥99% (HPLC)

form

DMSO solution

concentration

10 mM in DMSO

storage temp.

2-8°C

SMILES string

[n]1(c2[nH]c(n[c](c2nc1)=O)N)COC(CO)CO

InChI

1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)11-3-14(7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)

InChI key

IRSCQMHQWWYFCW-UHFFFAOYSA-N

Application

Ganciclovir ready-made solution has been used in combination with resveratrol to study its effect on melanoma cells. It has also been used as a synthetic substrate for viral thymidine kinase to study its effects on human-inducible pluripotent stem cells.

Biochem/physiol Actions

Ganciclovir is a nucleoside guanosine analog that contains an active moiety, ganciclovir triphosphate. This active moiety can inhibit viral replication by interrupting the viral DNA during elongation. Ganciclovir is a potent agent in preventing cytomegalovirus (CMV) retinitis disease in human immunodeficiency virus (HIV) patients. It exhibits therapeutic effects against CMV disease involving the lungs, liver, and gastrointestinal tract. Ganciclovir can be enzymatically phosphorylated to an active triphosphate analog by herpes simplex virus thymidine kinase (HSV-TK) or host-encoded deoxyguanosine kinase. It has the potential to treat allogeneic bone marrow, kidney, heart, liver, and lung transplantations.

Other Notes

Ganciclovir ready made solution in DMSO is provided at 10mM concentration (2.55mg/mL). It is suggested to dilute the provided ready made solution to a working concentration of 2-100μM for cell culture applications.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Tseng et al.
The Canadian journal of infectious diseases = Journal canadien des maladies infectieuses, 7(3), 183-194 (1996-05-01)
To review the pharmacology and pharmacokinetics of intravenous, oral and intraocular ganciclovir, and to discuss the role of these various formulations in the management of cytomegalovirus (CMV) retinitis in AIDS patients. A MEDLINE search (1987 through November 1995) of English-language
Targeting of herpes simplex virus 1 thymidine kinase gene sequences into the OCT4 locus of human induced pluripotent stem cells.
Ou, et al.
PLoS ONE, 8, e81131-e81131 (2015)
C J Oon et al.
Antiviral research, 41(3), 113-118 (1999-05-13)
Lamivudine is a new antiviral agent effective against hepatitis B viral (HBV) infections but can result in virus-drug resistance associated with mutations in the conserved 'YM552DD' motif of the HBV DNA polymerase. Due to their overlapping coding regions in the
J K McGavin et al.
Drugs, 61(8), 1153-1183 (2001-07-24)
Ganciclovir is a nucleoside guanosine analogue which incorporates ganciclovir triphosphate (the active moiety) into DNA during elongation, thereby inhibiting viral replication. Comparative studies of pre-emptive and prophylactic ganciclovir therapies in bone marrow transplant (BMT) recipients have shown similar rates of
Yuan Chen et al.
Journal of cancer research and clinical oncology, 146(6), 1489-1499 (2020-04-05)
To investigate the synergistic effect of resveratrol on the bystander effect of TK/GCV suicide gene system in melanoma cells. The effect of resveratrol on the growth of B16 cells and the synergistic effect of resveratrol with or without GCV were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service