Skip to Content
Merck
All Photos(1)

Key Documents

SML2179

Sigma-Aldrich

LDC000067 hydrochloride

≥98% (HPLC)

Synonym(s):

3-((6-(2-Methoxyphenyl)pyrimidin-4-yl)amino)phenyl)methanesulfonamide hydrochloride, 3-[[6-(2-Methoxyphenyl)-4-pyrimidinyl]amino]-benzenemethanesulfonamide, LDC067

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N4O3S · xHCl
CAS Number:
Molecular Weight:
370.43 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=S(CC1=CC(NC2=NC=NC(C3=CC=CC=C3OC)=C2)=CC=C1)(N)=O

InChI

1S/C18H18N4O3S/c1-25-17-8-3-2-7-15(17)16-10-18(21-12-20-16)22-14-6-4-5-13(9-14)11-26(19,23)24/h2-10,12H,11H2,1H3,(H2,19,23,24)(H,20,21,22)

InChI key

GGQCIOOSELPMBB-UHFFFAOYSA-N

Biochem/physiol Actions

LDC000067 (LDC067) is an ATP-competitive, potent and selective cyclin-dependent kinase 9 (CDK9) inhibitor (IC50 = 44 nM/CDK9-CycT1 vs. 5.51 μM/CDK1-CycB1, 2.44 μM/CDK2-CycA, 9.24 μM/CDK4-CycD1, >10 μM/CDK6-CycD3 & CDK7-CycH-MAT1; Kd = 32.7 nM/CDK9-CycT1 vs. 1.01 μM/CDK2-CycA, 16.0 μM/CDK7-CycH-MAT1) with much reduced or little potency against a panel of 28 non-CDK kinases. LDC067 selectively inhibits cellular RNA polymerase II CTD Ser2 phosphorylation (by 40% in HeLa; 60-min, 10 μM LDC067), but not CDK9-independent pSer5 or pSer7, causing apoptosis induction (by 239% and 200% of untreated control in A549 and MCF7 cultures) as a result of blocking RNAPII-mediated mRNA synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadine Löschmann et al.
Oncotarget, 7(36), 58051-58064 (2016-08-16)
The CDK inhibitor SNS-032 had previously exerted promising anti-neuroblastoma activity via CDK7 and 9 inhibition. ABCB1 expression was identified as major determinant of SNS-032 resistance. Here, we investigated the role of ABCB1 in acquired SNS-032 resistance. In contrast to ABCB1-expressing
Ke Ren et al.
PLoS pathogens, 12(10), e1005950-e1005950 (2016-10-21)
The human HSV-1 and -2 are common pathogens of human diseases. Both host and viral factors are involved in HSV lytic infection, although detailed mechanisms remain elusive. By screening a chemical library of epigenetic regulation, we identified bromodomain-containing protein 4
Xinli Qu et al.
Kidney international, 88(6), 1323-1335 (2015-07-30)
Transforming growth factor-β1 (TGF-β1)/Smad signaling has a central role in the pathogenesis of renal fibrosis. Smad3 and Smad4 are pro-fibrotic, while Smad2 is anti-fibrotic. However, these Smads form heterogeneous complexes, the functions of which are poorly understood. Here we studied
T K Albert et al.
British journal of pharmacology, 171(1), 55-68 (2013-10-10)
The cyclin-dependent kinase CDK9 is an important therapeutic target but currently available inhibitors exhibit low specificity and/or narrow therapeutic windows. Here we have used a new highly specific CDK9 inhibitor, LDC000067 to interrogate gene control mechanisms mediated by CDK9. The
Wanhua Xie et al.
Genome biology, 18(1), 32-32 (2017-02-18)
Monoubiquitination of H2B (H2Bub1) is a largely enigmatic histone modification that has been linked to transcriptional elongation. Because of this association, it has been commonly assumed that H2Bub1 is an exclusively positively acting histone modification and that increased H2Bub1 occupancy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service