Skip to Content
Merck
All Photos(1)

Key Documents

SML2163

Sigma-Aldrich

HPN-07

≥98% (HPLC)

Synonym(s):

2, 4-Disulfonyl PBN, 2,4-Disulfophenyl-N-tert-butylnitrone, ARL 16556, CPI 22, Cerovive, Disodium 4-[(tert-butyl-imino) methyl]benzene-13-disulfonate N-oxide, Disufenton sodium, NXY-059, Nxy 059, OKN-007, OKN007, Oklahoma nitrone-007

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO7S2 · 2Na
CAS Number:
Molecular Weight:
381.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[O-]/[N+](C(C)(C)C)=C\C(C=CC(S([O-])(=O)=O)=C1)=C1S([O-])(=O)=O.[Na+].[Na+]

InChI

1S/C11H15NO7S2.2Na/c1-11(2,3)12(13)7-8-4-5-9(20(14,15)16)6-10(8)21(17,18)19;;/h4-7H,1-3H3,(H,14,15,16)(H,17,18,19);;/q;2*+1/p-2

InChI key

XLZOVRYBVCMCGL-UHFFFAOYSA-L

Biochem/physiol Actions

Orally available NO-mimicking free radical trapping agent and sulfatase 2 (SULF2) inhibitor with in vivo efficacy in animal models of glioma and stroke.
Originally characterized for its NO-mimicking activity in preventing peroxynitrite formation and in vivo neuroprotective efficacy in animal models of cerebral ischemia (100 mg/kg i.v. in rabbits; 30-60 mg/kg plus 30-60 mg/kg/h i.v. or 50 mg/kg plus 8.8 mg/kg/h s.c. in rats; 28 mg/kg plus 16 mg/kg/h i.v. in monkeys), OKN-007 (HPN-07, NXY-059) is an orally available α-phenyl-tert-butylnitrone (PBN) derivative that is also shown to exhibit sulfatase 2 (SULF2) inhibitory activity and anticancer efficacy both in cultures (effective conc. 170-200 μM in Huh7 heptoma cultures) and in several rodent glioma models in vivo (C6, RG2, and GL261; 75 mg/kg/day for rats and 168 mg/kg/day for mice via drinking water).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Marshall et al.
Stroke, 32(1), 190-198 (2001-01-04)
NXY-059 is a novel nitrone with free radical-trapping properties that has a considerable neuroprotective effect in rats. We have now examined the efficacy of this drug at reducing long-term functional disability in a primate model of stroke. Twelve monkeys were
Z Zhao et al.
Brain research, 909(1-2), 46-50 (2001-08-02)
Free radicals have gained wide acceptance as mediators of cerebral ischemic injury. It has previously been reported that a spin trap nitrone, alpha-phenyl-N-tert-butyl nitrone (PBN), can reduce infarct volumes in rats subjected to either permanent or transient focal cerebral ischemia.
Philippe Garteiser et al.
Journal of magnetic resonance imaging : JMRI, 31(4), 796-806 (2010-04-08)
To demonstrate that OKN007, a disulfonyl derivative of phenyl-tert-butyl nitrone (PBN), has anti-glioma activity in the clinically relevant C6 rat glioma model using multi-parametric magnetic resonance imaging. Twenty-one rats were intracerebrally implanted with C6 cells and administered OKN007 or kept
Paul A Lapchak et al.
Stroke, 33(6), 1665-1670 (2002-06-08)
It has been proposed that the novel spin trap agent disodium-[(tert-butylimino)methyl]benzene-1,3-disulfonate N-oxide (NXY-059) may be useful in the treatment of ischemia and stroke. To date, there is little information concerning the safety of NXY-059 when administered in combination with the
Xin Zheng et al.
Genes, chromosomes & cancer, 52(3), 225-236 (2012-10-31)
Human sulfatase 2 (SULF2) functions as an oncoprotein in hepatocellular carcinoma (HCC) development by promoting tumor growth and metastasis via enhancement of fibroblast growth factor-2/extracellular signal-regulated kinase and WNT/β-catenin signaling. Recent results implicate that SULF2 activates the transforming growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service