Skip to Content
Merck
All Photos(1)

Key Documents

SML2090

Sigma-Aldrich

NAMI-A

≥98% ruthenium (Ru) basis (elemental analysis)

Synonym(s):

1H-Imidazole (OC-6-11)-tetrachloro(1H-imidazole-κN3)[1,1′-(sulfinyl-κS)bis[methane]]ruthenate(1-) (1:1), ImH[trans-RuCl4(DMSO)Im], Imidazolium trans-imidazoledimethyl sulfoxide-tetrachlororuthenate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10Cl4N2ORuS · C3H5N2
CAS Number:
Molecular Weight:
458.18
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% ruthenium (Ru) basis (elemental analysis)

form

powder

storage condition

desiccated

color

faint red to dark brown

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CS(C)=O.Cl[Ru](Cl)(Cl)Cl.N1=CNC=C1.C2=[NH+]C=CN2

Biochem/physiol Actions

NAMI-A is a potent antimetastatic drug in vivo that is exhibits little cytotoxicity towards many cancer cell lines. Antimetastatic of NAMI-A is attributed to the formation of adducts with membrane and cytosolic proteins. Nevertheless NAMI-A induces potent and selective cytotoxic effects in several leukemia cell lines. NAMI-A exhibits low toxicity for host tissues.
NAMI-A is a ruthenium-based anticancer agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Śpiewak et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 20(4), 695-703 (2015-03-21)
Imidazolium trans-tetrachloridodimethylsulfoxideimidazolruthenate(III), NAMI-A, a novel antimetastatic ruthenium complex was investigated towards affinity to transferrin (Tf), whether Tf-Ru adducts might be formed after its intravenous injection. Studies were focused on the holotransferrin due to its preferential binding to transferrin receptor. Here
Serena Pillozzi et al.
Dalton transactions (Cambridge, England : 2003), 43(32), 12150-12155 (2014-07-01)
We report here that the established anticancer ruthenium(iii) complex NAMI-A induces potent and selective cytotoxic effects in a few leukaemia cell lines. These results sound very surprising after 20 years of intense studies on NAMI-A, commonly considered as a "non-cytotoxic"
Alberta Bergamo et al.
Journal of inorganic biochemistry, 168, 90-97 (2017-01-09)
Solid tumours are constituted of tumour cells, healthy cells recruited from the host tissues and soluble factors released by both these cell types. The present investigation examines the capacity of co-cultures between the HCEC colon epithelial cells and the HCT-116
Metal-DNA coordination complexes
Pizarro AM, et al.
3D printing and additive manufacturing (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service