Skip to Content
Merck
All Photos(1)

Key Documents

SML2014

Sigma-Aldrich

Deracoxib

≥98% (HPLC)

Synonym(s):

4-[3-(Difluoromethyl)-5-(3-fluoro-4-methoxyphenyl)-1H-pyrazol-1-yl]benzenesulfonamide, 4-[5-(3-Fluoro-4-methoxyphenyl)-3-(difluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, SC-046, SC-46, SC-59046

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14F3N3O3S
CAS Number:
Molecular Weight:
397.37
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

FC(F)c1n[n](c(c1)c3cc(c(cc3)OC)F)c2ccc(cc2)[S](=O)(=O)N

InChI

1S/C17H14F3N3O3S/c1-26-16-7-2-10(8-13(16)18)15-9-14(17(19)20)22-23(15)11-3-5-12(6-4-11)27(21,24)25/h2-9,17H,1H3,(H2,21,24,25)

InChI key

WAZQAZKAZLXFMK-UHFFFAOYSA-N

Biochem/physiol Actions

Deracoxib is an orally active cyclooxygenase-2 (COX-2) inhibitor with >48-fold selectivity over COX-1 (IC50 of 203-189 nM against COX-2-dependent PGE2 production vs. IC50 of 9.85-9.96 μM against COX-1-dependent TxB2 production in dog whole blood). Deracoxib is also reported to exhibit inhibitory activity against phosphodiesterase (Ki = 3.6 μM against human PDE4D3) as well as positive modulatory effiacy toward 2-APB-stimulated transient receptor potential vanilloid 3 channel (TRPV3) activity.
Orally active COX-2 inhibitor with >48-fold selectivity over COX-1. Also inhibits PDE4D3 and enhances 2-APB-stimulated TRPV3 activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J N King et al.
Research in veterinary science, 88(3), 497-506 (2009-12-17)
In vitro whole blood canine assays were used to quantify the inhibitory actions of the novel non-steroidal anti-inflammatory drug (NSAID) robenacoxib on the cyclooxygenase (COX) isoenzymes, COX-1 and COX-2, in comparison with other drugs of the NSAID class. COX-1 activity
Jacques Pantel et al.
European journal of pharmacology, 660(1), 139-147 (2011-02-08)
The melanocortin MC(4) receptor is a potential target for the development of drugs for both obesity and cachexia. Melanocortin MC(4) receptor ligands known thus far are orthosteric agonists or antagonists, however the agonists, in particular, have generally exhibited unwanted side
Stefan Spyra et al.
British journal of pharmacology, 174(16), 2696-2705 (2017-06-02)
The transient receptor potential vanilloid 3 (TRPV3) channel is a heat-sensitive ion channel, which is predominantly expressed in keratinocytes. TRPV3 channels are involved in numerous physiological and pathophysiological processes within the skin, including cutaneous nociception, temperature sensation and development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service