Skip to Content
Merck
All Photos(1)

Key Documents

SML1914

Sigma-Aldrich

TUG-891

≥98% (HPLC)

Synonym(s):

3-(4-((4-Fluoro-4-methyl-[1,1-biphenyl]-2-yl)methoxy)phenyl)-propanoic acid, 4-[(4-Fluoro-4′-methyl[1,1′-biphenyl]-2-yl)methoxy]-benzenepropanoic acid, TUG 891, TUG891

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21FO3
CAS Number:
Molecular Weight:
364.41
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

Fc1cc(c(cc1)c3ccc(cc3)C)COc2ccc(cc2)CCC(=O)O

InChI

1S/C23H21FO3/c1-16-2-7-18(8-3-16)22-12-9-20(24)14-19(22)15-27-21-10-4-17(5-11-21)6-13-23(25)26/h2-5,7-12,14H,6,13,15H2,1H3,(H,25,26)

InChI key

LPGBXHWIQNZEJB-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, potent and selective agonist for long chain free fatty acid (FFA) receptor FFA4 (GPR120).
TUG-891 is an orally active, potent and selective agonist for human long chain free fatty acid (FFA) receptor FFA4/GPR120 (pEC50 of receptor β-arrestin-2 recruitment = 7.36/FFA4 vs. 4.19/FFA1 HEK transfectants; pEC50 of calcium mobilization = 7.02/FFA4 vs. 5.30/FFA1 transfectants), while exhibiting no activity toward human short-chain FFA receptors FFA2/GPR43 and FFA3/GPR41. TUG-891 also exhibits potent partial agonist activity toward murine FFA4 (pEC50 = 7.77, 75% efficacy by β-arrestin-2 recruitment assay; pEC50 = 6.89 by calcium mobilization), albeit with a reduced selectivity over mFFA1 (pEC50 = 5.92 by β-arrestin-2 recruitment and 6.41 by by calcium mobilization). When administered via water intake (~20 mg/kg/day), TUG-891 is reported to reverse food intake increases and body weight gains among mice subjected to sleep fragmentation, and significantly attenuate visceral white adipose tissue inflammation as well as insulin resistance.
TUG-891, a GPR120 agonist helps to stimulate metabolic health by inducing mitochondrial respiration in brown fat. It is capable of enhancing lipid oxidation and decreasing fat mass in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Gozal et al.
International journal of obesity (2005), 40(7), 1143-1149 (2016-03-17)
Sleep fragmentation (SF), a frequent occurrence in multiple sleep and other diseases leads to increased food intake and insulin resistance via increased macrophage activation and inflammation in visceral white adipose tissue (VWAT). Free fatty acid receptor 4 (FFA4) is reduced
Brian D Hudson et al.
Molecular pharmacology, 84(5), 710-725 (2013-08-28)
TUG-891 [3-(4-((4-fluoro-4'-methyl-[1,1'-biphenyl]-2-yl)methoxy)phenyl)propanoic acid] was recently described as a potent and selective agonist for the long chain free fatty acid (LCFA) receptor 4 (FFA4; previously G protein-coupled receptor 120, or GPR120). Herein, we have used TUG-891 to further define the function
The GPR120 agonist TUG-891 promotes metabolic health by stimulating mitochondrial respiration in brown fat
Schilperoort M, et al.
EMBO Molecular Medicine, 10(3) (2018)
Bharat Shimpukade et al.
Journal of medicinal chemistry, 55(9), 4511-4515 (2012-04-24)
GPR120 is a receptor of unsaturated long-chain fatty acids reported to mediate GLP-1 secretion, insulin sensitization, anti-inflammatory, and anti-obesity effects and is therefore emerging as a new potential target for treatment of type 2 diabetes and metabolic diseases. Further investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service