SML1687
3BDO
≥98% (HPLC)
Synonym(s):
3-Benzyl-5-((2-nitrophenoxy)methyl)-dihydrofuran-2(3H)-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
oil
color
colorless to light brown
storage temp.
2-8°C
SMILES string
O=C1OC(COC2=C([N+]([O-])=O)C=CC=C2)CC1CC3=CC=CC=C3
Application
3BDO has been used to investigate the potential effects of metformin on inflammatory lung injury.
Biochem/physiol Actions
3BDO is a cell-permeable, orally bioavailable, non-toxic butyrolactone derivative that is shown to competitively bind FKBP1A (FK506-binding protein 1A, 12 kDa) in a reversible manner, and suppress autophagy via the mTOR pathway. 3BDO inhibits both LPS- and oxLDL-induced autophagy in HUVECs, increases TIA1 phosphorylation, and reduces autophagosomes number. It is reported to be brain permeant, and significantly decreases atherosclerosis development in apoE-/- mice (100 mg/kg, q.d., p.o.). 3BDO increases IDE and neprilysin levels, lowers amyloid-β deposition in an AβPP/PS1 transgenic mouse model of Alzheimer′s disease, and alleviates memory deficits.
An orally bioavailable, brain-permeant suppressor of mTOR-mediated autophagy; reduces Aβ levels and prevents cognitive deficits in AβPP/PS1 mouse model.
3-benzyl-5-[(2-nitrophenoxy) methyl]-dihydrofuran-2 (3H)-one (3BDO) blocks rise in LC3-II stimulated by uric acid (UA). It protects the capability of Rheb1-deficient macrophages to perform phagocytosis. 3BDO also prevents the inhibitory effects of EX-527 on eukaryotic translation initiation factor-binding protein 1(4E-BP1) phosphorylation.
3-benzyl-5-[(2-nitrophenoxy) methyl]-dihydrofuran-2 (3H)-one (3BDO) blocks rise in LC3-II stimulated by uric acid (UA). It protects the capability of Rheb1-deficient macrophages to perform phagocytosis. 3BDO also prevents the inhibitory effects of EX-527 on eukaryotic translation initiation factor-binding protein 1(4E-BP1) phosphorylation.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Experimental neurology, 297, 138-147 (2017-08-20)
Serum urate levels are reported to be significantly lowered in patients with Parkinson's disease (PD) and inversely correlated to the risk and progression of PD. However, the mechanism by which urate affects PD is poorly understood. Here we showed that
The SIRT1 inhibitor EX-527 suppresses mTOR activation and alleviates acute lung injury in mice with endotoxiemia.
Innate Immunity, 23(8), 678-686 (2017)
Rheb1-mTORC1 maintains macrophage differentiation and phagocytosis in mice.
Experimental Cell Research, 344(2), 219-228 (2016)
Metformin alleviated endotoxemia-induced acute lung injury via restoring AMPK-dependent suppression of mTOR.
Chemico-Biological Interactions, 291, 1-6 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service