Skip to Content
Merck
All Photos(1)

Key Documents

SML1496

Sigma-Aldrich

Buformin hydrochloride

≥98% (HPLC)

Synonym(s):

1-Butylbiguanide hydrochloride, N-Butyl-imidodicarbonimidic diamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N5 · HCl
CAS Number:
Molecular Weight:
193.68
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

−20°C

InChI

1S/C6H15N5.ClH/c1-2-3-4-10-6(9)11-5(7)8;/h2-4H2,1H3,(H6,7,8,9,10,11);1H

InChI key

KKLWSPPIRBIEOV-UHFFFAOYSA-N

Application

Buformin hydrochloride has been used as an internal standard in liquid chromatography-tandem mass spectrometry to analyze postmortem distribution/redistribution of buformin in body fluids and solid tissues in an autopsy case.

Biochem/physiol Actions

Buformin is an oral antidiabetic drug with antiproliferative properties; AMPK activator. Buformin, an antidiabetic biguanide drug, was withdrawn as an antidiabetic drug in several countries because of the side effect of lactic acidosis, but is now being investigated as an anticancer agent. Buformin inhibits the growth and development of cancer. Buformin has been shown to postpone spontaneous carcinogenesis as well as inhibit induced carcinogenesis in animal studies. Buformin decreased cancer incidence, multiplicity, and burden in a rat study of chemically induced mammary carcinogenesis. Buformin also reduced renal cell carcinoma survival under conditions of glucose deprivation. Buformin′s anticancer activity is thought to involve disruption of the Warburg effect by AMPK activation, which decreases hepatic glucose production, and inhibition of the mTor pathway.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service