Skip to Content
Merck
All Photos(1)

Key Documents

SML1421

Sigma-Aldrich

EPZ015666

≥98% (HPLC)

Synonym(s):

(S)-N-(3-(3,4-Dihydroisoquinolin-2(1H)-yl)-2-hydroxypropyl)-6-(oxetan-3-ylamino)pyrimidine-4-carboxamide, GSK3235025, YQ36286

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N5O3
CAS Number:
Molecular Weight:
383.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@H](CN1CCC(C=CC=C2)=C2C1)CNC(C3=CC(NC4COC4)=NC=N3)=O

InChI

1S/C20H25N5O3/c26-17(10-25-6-5-14-3-1-2-4-15(14)9-25)8-21-20(27)18-7-19(23-13-22-18)24-16-11-28-12-16/h1-4,7,13,16-17,26H,5-6,8-12H2,(H,21,27)(H,22,23,24)/t17-/m0/s1

InChI key

ZKXZLIFRWWKZRY-KRWDZBQOSA-N

Biochem/physiol Actions

EPZ015666 (GSK3235025) is a protein arginine methyltransferase PRMT5 inhibitor with an IC50 value of of 22 nM in biochemical assays and no inhibition up to the maximum tested concentration of 50 μM against a panel of 20 other protein methyltransferases. Inhibition was uncompetitive with respect to SAM. EPZ015666 showed dose-dependent antitumor activity in multiple in vitro and in vivo mantle cell lymphoma (MCL) models with treatment of MCL cell lines leading to inhibition of SmD3 methylation and cell death.

Other Notes

EPZ015666 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the EPZ015666 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosemary Millen et al.
Med (New York, N.Y.), 4(5), 290-310 (2023-05-14)
Organoids are in vitro three-dimensional structures that can be grown from patient tissue. Head and neck cancer (HNC) is a collective term used for multiple tumor types including squamous cell carcinomas and salivary gland adenocarcinomas. Organoids were established from HNC patient
Zhengxin Wang et al.
Molecular and cellular biology, 42(4), e0052321-e0052321 (2022-03-22)
Protein arginine methyltransferase (PRMT) enzymes catalyze posttranslational modifications of target proteins and are often upregulated in human cancers. In this study, we purified two chemical compounds from seeds of Foeniculum vulgare based on their ability to inhibit the enzymatic activity
David Shifteh et al.
Cancers, 12(8) (2020-08-01)
Nearly 45% of colorectal cancer (CRC) patients harbor a mutation in their KRAS gene for which, despite many years of research, there are still no targeted therapies available. Protein Arginine Methyltransferase 5 (PRMT5) is a transcription regulator for multiple cellular
Pan-methylarginine antibody generation using PEG linked GAR motifs as antigens.
Wang, et al.
Methods, 200, 80-86 (2023)
Kazuki Fukumoto et al.
Communications biology, 5(1), 313-313 (2022-04-07)
The global dietary supplement market is valued at over USD 100 billion. One popular dietary supplement, S-adenosylmethionine, is marketed to improve joints, liver health and emotional well-being in the US since 1999, and has been a prescription drug in Europe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service