Skip to Content
Merck
All Photos(1)

Key Documents

SML1334

Sigma-Aldrich

WH-4-023

≥98% (HPLC)

Synonym(s):

2,6-Dimethylphenyl N-(2,4-dimethoxyphenyl)-N-[2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]carbamate, KIN112

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N6O4
CAS Number:
Molecular Weight:
568.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CN1CCN(C2=CC=C(NC3=NC=CC(N(C4=CC=C(OC)C=C4OC)C(OC5=C(C)C=CC=C5C)=O)=N3)C=C2)CC1

InChI

1S/C32H36N6O4/c1-22-7-6-8-23(2)30(22)42-32(39)38(27-14-13-26(40-4)21-28(27)41-5)29-15-16-33-31(35-29)34-24-9-11-25(12-10-24)37-19-17-36(3)18-20-37/h6-16,21H,17-20H2,1-5H3,(H,33,34,35)

InChI key

NBTNHSGBRGTFJS-UHFFFAOYSA-N

Biochem/physiol Actions

WH-4-023 (KIN112) is a potent and selective inihbitor of the tyrosine kinases Lck and Src with IC50 values of 2 nM for Lck and 6 nM for Src. WH-4-023 is a somewhat less potent inhibitor of salt-inducible kinases (SIKs) with IC50 values of 10, 22 and 60 nM for SIK 1, 2 and 3 respectively. WH-4-023 was found to support self-renewal of naive human embryonic stem cells, in particular in combination with PD 0325901 (PZ0162), CHIR99021 (SML1046), and SB-590885 (SML0501).
WH-4-023 belongs to the Src kinase family. It stimulates apoptosis in GC-resistant T-acute lymphoblastic leukemia (T-ALL) cells.

Other Notes

WH-4-023 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the WH-4-023 probe summary on the Chemical Probes Portal website.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting loss of the Hippo signaling pathway in NF2-deficient papillary kidney cancers
Sourbier, et al.
Oncotarget, 9(12), 10723-10723 (2018)
Glucocorticoid resistance is reverted by LCK inhibition in pediatric T-cell acute lymphoblastic leukemia
Serafin, et al.
Blood, blood-b2017 (2017)
Glucocorticoid resistance is reverted by LCK inhibition in pediatric T-cell acute lymphoblastic leukemia
Serafin, et al.
Blood, blood-b2017 (2017)
Cheong-Yong Yun et al.
Theranostics, 9(3), 646-660 (2019-02-28)
Rationale: SOX10 (SRY-related HMG-box 10) and MITF-M (microphthalmia-associated transcription factor M) restrict the expression of melanogenic genes, such as TYR (tyrosinase), in melanocytes. DACE (diacetylcaffeic acid cyclohexyl ester) inhibits melanin production in α-MSH (α-melanocyte stimulating hormone)-activated B16-F0 melanoma cells. In

Articles

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service