Skip to Content
Merck
All Photos(1)

Key Documents

SML1120

Sigma-Aldrich

Thiophene-2

≥98% (HPLC)

Synonym(s):

Methyl 2-(perfluorobenzamido)-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, TP2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14F5NO3S
CAS Number:
Molecular Weight:
419.37
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC)C1=C(NC(C2=C(F)C(F)=C(F)C(F)=C2F)=O)SC3=C1CCCCC3

InChI

1S/C18H14F5NO3S/c1-27-18(26)9-7-5-3-2-4-6-8(7)28-17(9)24-16(25)10-11(19)13(21)15(23)14(22)12(10)20/h2-6H2,1H3,(H,24,25)

InChI key

AVRWEULSKHQETA-UHFFFAOYSA-N

Biochem/physiol Actions

Thiophene-2 (TP2) is an inhibitor of polyketide synthase 13 (Pks13), which plays a critical role in the biosynthesis of mycolic acid, an essential component of the cell wall in M. tuberculosis, and is a potential new target for tuberculosis treatment. TP2 inhibits FadD32-dependent loading of the mycolic acid precursor meromycoloyl chain onto acyl carrier protein (ACP) domains located at the N terminus of Pks13, preventing synthesis of mycolic acid and resulting in mycobacterial cell death. TP2 bacteriosidal activity is equivalent to treatment with the first-line drug isoniazid, and enhances its activity, but is less likely to cause resistance. The minimal inhibitory concentration (MIC) values of TP2 against drug-susceptible multidrug-resistant M. tuberculosis strains is approximately 1 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongmin Zhao et al.
Veterinary microbiology, 245, 108708-108708 (2020-05-28)
The outbreak and spread of Tembusu virus (TMUV) has caused very large losses in the waterfowl-breeding industry since 2010. The viral envelope (E) protein, the principal surface protein of viral particles, plays a vital role in viral entry and fusion.
Katherine Sorber et al.
Nucleic acids research, 39(9), 3820-3835 (2011-01-20)
Over 50% of genes in Plasmodium falciparum, the deadliest human malaria parasite, contain predicted introns, yet experimental characterization of splicing in this organism remains incomplete. We present here a transcriptome-wide characterization of intraerythrocytic splicing events, as captured by RNA-Seq data
"
Valentina Serra et al.
Frontiers in microbiology, 7, 1704-1704 (2016-11-22)
Bihong T Chen et al.
Breast cancer research and treatment, 176(1), 181-189 (2019-04-17)
Older cancer patients are at increased risk of cancer-related cognitive impairment. The purpose of this study was to assess the alterations in intrinsic brain activity associated with adjuvant chemotherapy in older women with breast cancer. Chemotherapy treatment (CT) group included
Richard E Morton et al.
Journal of lipid research, 61(8), 1168-1179 (2020-06-28)
Cholesteryl ester transfer protein (CETP) facilitates the net transfer of cholesteryl esters (CEs) and TGs between lipoproteins, impacting the metabolic fate of these lipoproteins. Previous studies have shown that a CETP antibody can alter CETP's preference for CE versus TG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service