Skip to Content
Merck
All Photos(1)

Key Documents

SML1109

Sigma-Aldrich

KU-55933

≥98% (HPLC)

Synonym(s):

2-(4-Morpholinyl)-6-(1-thianthrenyl)-4H-Pyran-4-one, 2-(Morpholin-4-yl)-6-(thianthren-1-yl)-4H-pyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17NO3S2
CAS Number:
Molecular Weight:
395.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C=C(OC(=C1)c2cccc3Sc4ccccc4Sc23)N5CCOCC5

InChI

1S/C21H17NO3S2/c23-14-12-16(25-20(13-14)22-8-10-24-11-9-22)15-4-3-7-19-21(15)27-18-6-2-1-5-17(18)26-19/h1-7,12-13H,8-11H2

InChI key

XRKYMMUGXMWDAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

KU-55933 has been used as an ataxia-telangiectasia mutated (Atm) inhibitor.

Biochem/physiol Actions

KU-55933 is a very potent, specific inhibitor of Ataxia telangiectasia (A-T) mutated (ATM) kinase (IC50 = 13 nM). KU-22933 treatment sensitizes cancer cells to ionizing radiation and cytotoxic drugs. The compound KU-22933 blocks ATM-mediated phosphorylyation of p53, gH2AX, NBS1, and SMC1.
KU-55933 is a very potent, specific inhibitor of Ataxia telangiectasia (A-T) mutated (ATM) kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christianne Hoey et al.
Molecular oncology, 12(8), 1324-1341 (2018-05-31)
Recurrence of high-grade prostate cancer after radiotherapy is a significant clinical problem, resulting in increased morbidity and reduced patient survival. The molecular mechanisms of radiation resistance are being elucidated through the study of microRNA (miR) that negatively regulate gene expression.
Zhongkai Zhang et al.
Journal of experimental & clinical cancer research : CR, 39(1), 247-247 (2020-11-18)
SIRT6 has diverse roles in cells, and the role of SIRT6 in tumorigenesis is controversial. Considering the role of SIRT6 as an inducer of DNA damage repair, it might be involved in resistance to anti-cancer therapy. We evaluated the prognostic
A surveillance mechanism ensures repair of DNA lesions during zygotic reprogramming.
Ladstatter S and Kikue T K
Cell, 167(7), 1774-1787 (2016)
Sanket Awate et al.
BMC molecular biology, 17, 3-3 (2016-02-11)
The Tousled like kinase 1B (TLK1B) is critical for DNA repair and survival of cells. Upon DNA damage, Chk1 phosphorylates TLK1B at S457 leading to its transient inhibition. Once TLK1B regains its kinase activity it phosphorylates Rad9 at S328. In
Bo-Hua Chen et al.
Nature communications, 11(1), 5267-5267 (2020-10-21)
Peroxisomes perform beta-oxidation of branched and very-long chain fatty acids, which leads to the formation of reactive oxygen species (ROS) within the peroxisomal lumen. Peroxisomes are therefore prone to ROS-mediated damages. Here, using light to specifically and acutely induce ROS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service