Skip to Content
Merck
All Photos(1)

Key Documents

SML1094

Sigma-Aldrich

CHIR 98014

≥98% (HPLC), powder, GSK-3 inhibitor

Synonym(s):

N-6-[2-[[4-(2,4-Dichlorophenyl)-5-(1H-imidazol-1-yl)-2-pyrimidinyl]amino]ethyl]-3-nitro-2,6-pyridinediamine, CHIR-98014

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17Cl2N9O2
CAS Number:
Molecular Weight:
486.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

CHIR 98014, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 0.5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

NC1=C(C=CC(NCCNC2=NC(C3=C(Cl)C=C(Cl)C=C3)=C(N4C=CN=C4)C=N2)=N1)[N+]([O-])=O

InChI

1S/C20H17Cl2N9O2/c21-12-1-2-13(14(22)9-12)18-16(30-8-7-24-11-30)10-27-20(29-18)26-6-5-25-17-4-3-15(31(32)33)19(23)28-17/h1-4,7-11H,5-6H2,(H3,23,25,28)(H,26,27,29)

InChI key

MDZCSIDIPDZWKL-UHFFFAOYSA-N

Related Categories

Application

CHIR 98014 has been used for the generation of small molecules neural progenitor cells and differentiation towards motor neurons. It has also been used as a Wnt/β-catenin pharmacological agonist in the cell-conditioned medium to perform chromatin immunoprecipitation (ChIP) studies in HT22 neurons.

Biochem/physiol Actions

CHIR 98014 is a glycogen synthase kinase-3 (GSK-3) inhibitor with IC50 values of 0.65 nM and 0.58 nM for GSK3α and GSK3β, respectively. CHIR98014 is 500-fold to >10,000-fold selectivity for GSK-3 versus 20 other protein kinases tested, including closest homologs Cdc2 and ERK2.
CHIR 98014 is a glycogen synthase kinase-3 (GSK-3) inhibitor.

Reconstitution

Reconstitute in DMSO at 0.5 mg/mL. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Storage and Stability

The product in the original sealed container is quite stable. It should be stored at −20°C in the powdered format.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matías A Medina et al.
Translational psychiatry, 8(1), 45-45 (2018-03-06)
Synaptic abnormalities have been described in individuals with autism spectrum disorders (ASD). The cell-adhesion molecule Neuroligin-3 (Nlgn3) has an essential role in the function and maturation of synapses and NLGN3 ASD-associated mutations disrupt hippocampal and cortical function. Here we show
Diana Gumber et al.
eLife, 9 (2020-12-18)
WNT proteins are secreted symmetry breaking signals that interact with cell surface receptors of the FZD family to regulate a multitude of developmental processes. Studying selectivity between WNTs and FZDs has been hampered by the paucity of purified WNT proteins
Paola Rusmini et al.
Autophagy, 15(4), 631-651 (2018-10-20)
Macroautophagy/autophagy, a defense mechanism against aberrant stresses, in neurons counteracts aggregate-prone misfolded protein toxicity. Autophagy induction might be beneficial in neurodegenerative diseases (NDs). The natural compound trehalose promotes autophagy via TFEB (transcription factor EB), ameliorating disease phenotype in multiple ND

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service