Skip to Content
Merck
All Photos(1)

Key Documents

SML1057

Sigma-Aldrich

NPP

≥98% (HPLC)

Synonym(s):

3-(4-Nitrophenyl)propiolic acid methyl ester, Methyl (4-nitrophenyl)-2-propynoate, Methyl 3-(4-nitrophenyl) propiolate, Methyl 3-(4-nitrophenyl)prop-2-ynoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO4
CAS Number:
Molecular Weight:
205.17
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C10H7NO4/c1-15-10(12)7-4-8-2-5-9(6-3-8)11(13)14/h2-3,5-6H,1H3

InChI key

XNRRUKAHRUCWGC-UHFFFAOYSA-N

Biochem/physiol Actions

NPP is a ROS promoter selective for cancer tissue. NPP is a potent and preferential tumor cell death inducer that causes apoptosis in cancer cells by cytochrome P450 catalyzed ROS formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service