Skip to Content
Merck
All Photos(1)

Key Documents

SML0670

Sigma-Aldrich

Leonurine

≥98% (HPLC)

Synonym(s):

4-Guanidino-1-butanol syringate, 4-Guanidinobutyl 4-hydroxy-3,5-dimethoxybenzoate, SCM-198

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O5
CAS Number:
Molecular Weight:
311.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

NC(=NCCCCOC(=O)c1cc(c(c(c1)OC)O)OC)N

InChI

1S/C14H21N3O5/c1-20-10-7-9(8-11(21-2)12(10)18)13(19)22-6-4-3-5-17-14(15)16/h7-8,18H,3-6H2,1-2H3,(H4,15,16,17)

InChI key

WNGSUWLDMZFYNZ-UHFFFAOYSA-N

Biochem/physiol Actions

Leonurine (SCM-198) is a natural product with antioxidant, anti-inflammatory, and cardioprotective activity being investigated for the treatment of cardiovascular disease and stroke. Leonurine suppresses inflammation and oxidative stress, decreasing a number of inflammatory factors, reducing cardiac Nox4 expression, ROS production, NF-KB activation, and plasma MMP-2 activity and increasing superoxide dismutase (SOD).
Leonurine is an antioxidant, anti-inflammatory cardioprotective agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

XinHua Liu et al.
European journal of pharmacology, 649(1-3), 236-241 (2010-09-22)
Leonurine, an alkaloid typically found in Herba leonuri, is known to have both antioxidant and cardioprotective properties. In the present study, we investigated the cardioprotective mechanism of leonurine the in vivo rat model of chronic myocardial ischemia and in vitro
Xin Hua Liu et al.
European journal of pharmacology, 680(1-3), 108-114 (2012-02-07)
Leonurine, an active alkaloid of Traditional Chinese Medicine Herba leonuri, displayed cardioprotective effects by anti-oxidative and anti-apoptotic activities in vitro and in vivo. Herein, we explored the effects and possible mechanisms of leonurine on lipopolysaccharide (LPS)-induced inflammatory responses in human
Bo Hou et al.
Folia neuropathologica, 60(1), 76-91 (2022-04-02)
Atherosclerosis (AS) is a chronic inflammatory disease with high morbidity and mortality worldwide and is the pathologic basis of cerebral-cardiovascular disease. Cerebral infarction is caused by cerebral ischemia in the deep brain stem or cerebral hemisphere, and small vessel atherosclerosis
Hui Huang et al.
Bioscience reports, 32(2), 185-195 (2011-08-24)
There are reports of early evidence that suggest the involvement of chronic low-grade inflammation in the pathogenesis of Type 2 diabetes. Thus, substances that have effects in reducing inflammation could be potential drugs for Type 2 diabetes. Leonurine (4-guanidino-n-butyl syringate; SCM-198) is
Kok Poh Loh et al.
Stroke, 41(11), 2661-2668 (2010-10-16)
Oxidative stress is known to be involved in ischemic stroke. Intense interest is drawn to the therapeutic potential of Chinese herbs on ischemic stroke because many of them contain antioxidant properties. Leonurine, 1 of the active compounds from purified Herba

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service